VWR Collection Catalogue - page 242
228
Temperature Treatment and Drying
Circulating Thermostats
Continued from previous page
Type
Cooling capacity (kW)
Pk
Cat. No.
1175PD
1,41/0,91/0,62*
1
1177MD
2,07/1,41/0,63*
1
1177PD
2,07/1,41/0,63*
1
1179MD
2,40/1,59/0,83*
1
1179PD
2,40/1,59/0,83*
1
* Cooling capacity at +20, +10 and 0 °C
Description
Pk
Cat. No.
Shaking water bath, VKB 12, EU-plug
1
Shaking water bath, VKB 12, UK-plug
1
Shaking water bath, VKB 18, EU-plug
1
Shaking water bath, VKB 18, UK-plug
1
Description
For
Pk
Cat. No.
Trays for VKB shaking water baths
Plain stainless steel tray for VKB 12
Containers, bags and other vessels
1
Plain stainless steel tray for VKB 18
Containers, bags and other vessels
1
Universal flask tray with springs, stainless steel for VKB 18
25×25 ml, 16×50 ml, 9×100 ml, 5×250 ml, 4×500 ml or 2×1000
1
Universal flask tray with springs, stainless steel for VKB 18
45×25 ml, 28×50 ml, 18×100 ml, 11×250 ml, 6×500 ml or 3×1000
1
Test tube tray, stainless steel for VKB 12
3 test tube racks
1
Test tube tray, stainless steel for VKB 18
5 test tube racks
1
Racks for VKB shaking water baths
Stainless steel test tube rack for VKB 12/VKB 18*
119×0,5 ml microtubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
48×1,5 ml microtubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
48×10 mm Ø tubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
44×13 mm Ø tubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
24×16 mm Ø tubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
21×19 mm Ø tubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
12×25 mm Ø tubes
1
Stainless steel test tube rack for VKB 12/VKB 18*
10×30 mm Ø tubes
1
Lids for VKB shaking water baths
Gabled stainless steel lid
VKB 12
1
Gabled stainless steel lid
VKB 18
1
Replacement gabled non drip polycarbonate lid
VKB 12
1
Replacement gabled non drip polycarbonate lid
VKB 18
1
Model
VKB 12
VKB 18
Flask immersion depth (mm)
0/60 min/max
Linear stroke length (mm)
18
Shaker tray W×D (mm)
240×235
420×235
Uniformity (°C)
±0,1 (at 37 °C)
Weight (kg)
10
13
W×D×H (mm)
335×390×270
335×570×270
* Note the tray racks must be used in conjunction with the appropriate stainless steel tray.
Shaking water baths, VKB series
Linear shaking water baths with stainless steel tank and simple, intuitive 3 button user interface. It is
quick and easy to fit alterative shaking trays. Heating element is positioned on the underside of tank, no
heating element in the bath means it is easier to clean and keep clean. Baths have an independent sample
protection feature. Available ready to use in two sizes to suit a variety of applications including sample
preparation, mixing and thawing.
Ş Intuitive user interface enabling quick changes to temperature and shaking speed
Ş Discrete, under tray, magnetically coupled shaking mechanism is reliable and quiet in operation
Ş Excellent temperature stability ±0,2°C
Ş Temperature range: Ambient + 5 to 99°C
Ş Adjustable shaking speed range: 40 to 400 strokes per minute (dependant on load)
Ş Display: LED, resolution 0,1°C
Delivery Information:
Supplied with a universal shaking tray and clear polycarbonate lid. A gabled lid is
recommended when working at temperatures above 60 °C, a wide choice of accessory trays and test tube
racks are available, these must be ordered separately.
1...,232,233,234,235,236,237,238,239,240,241 243,244,245,246,247,248,249,250,251,252,...612
Powered by FlippingBook