VWR Collection Catalogue - page 240
226
Temperature Treatment and Drying
Circulating Thermostats
Type
Temp. range
(°C)
Temp. stability
(°C)
Heating
capacity (kW)
Cooling capacity
(kW)
Capacity (l)
Bath W×D×H
(mm)
W×D×H (mm)
Pk
Cat. No.
Models with Advanced Digital temperature controller
AD7LR-20
-20…+200
±0,01
2,2
0,2
7
142×157×127 411×589×411
1
AD07R-20
-20…+200
±0,01
2,2
0,2
7
142×157×127 221×541×617
1
AD07R-40
-40…+200
±0,01
2,2
0,505
7
142×157×127 221×541×617
1
AD15R-30
-30…+200
±0,01
2,2
0,915
15
276×212×140 368×569×683
1
AD15R-40
-40…+200
±0,01
2,2
1,0
15
276×212×140 368×569×683
1
AD20R-30
-30…+200
±0,01
2,2
0,915
20
316×250×140 419×610×683
1
AD28R-30
-30…+200
±0,01
2,2
0,915
28
359×314×140 457×672×683
1
AD45R-20
-25…+135
±0,01
2,2
1,4
45
398×549×140 568×909×1070
1
Models with Advanced Programmable temperature controller
AP07R-20
-20...+200
±0,01
2,2
0,2
7
142×157×127 221×541×617
1
AP07R-40
-40…+200
±0,01
2,2
0,505
7
142×157×127 221×541×617
1
AP15R-30
-30…+200
±0,01
2,2
0,915
15
276×212×140 368×569×683
1
AP15R-40
-40…+200
±0,01
2,2
1,0
15
276×212×140 368×569×683
1
AP20R-30
-30…+200
±0,01
2,2
0,915
20
316×250×140 419×610×683
1
AP28R-30
-30…+200
±0,01
2,2
0,915
28
359×314×140 457×672×683
1
AP45R-20
-25…+135
±0,01
2,2
1,4
45
398×549×140 568×909×1070
1
Note: Cooling capacity at 20 °C
Refrigerated circulators
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both
are easy to navigate with large, intuitive displays and multiple communication options including USB-A & B, RS232/485,
Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models
feature user-adjustable high temperature safety cut-off points as well as over-temperature protection. The DuraTop™
surface is cool to the touch when operating at high temperatures.
Digital
models feature an intuitive 9,5 cm display with touch pad control, single point calibration, menus and prompts in
4 languages: French, German, Spanish and English.
Programmable
models have an intuitive 10,9 cm SmartTouch display, 5-point calibration capability, menus and prompts
in 6 languages: French, German, Spanish, English, Chinese and Arabic, plus time/temperature programming (ten 100-
step programmes).
ŞWorking temperatures from -40 to +200°C with stability of ±0.01 °C
ŞVariable-speed pressure/suction pump with external circulation and temperature control capability
ŞSwivel 180™ rotating controllers, LidDock™ lid stowing system, DuraTop™ chemically resistant deck and
WhisperCool™ environmental control system reduces noise, increases refrigeration efficiency and lowers energy
consumption
Ş Cool Command™ technology regulates the amount of cooling required, saving energy while providing rapid cooling and precise control at elevated
temperatures
Ş Event scheduling (time & date), real-time clock and temperature trends for up to 10 days with Programmable models
Ş Selectable home screens and on-screen help
Ş Automatic and/or user-adjustable performance optimisation
Comply with DIN 12876-1, Safety class III
Maximum pump capacities:
16.7 l/min, 250 mbar, 12.2 l/min suction
Delivery Information:
Supplied with a reservoir cover, bypass tubing, male inlet and outlet adapters for 47, 63 and 95 mm tubing, ¼” to M16 adapters are also
included. For probes and accessories, please visit vwr.com or contact our Technical Office on 01342 337447.
Description
Pk
Cat. No.
High temperature tubing
1
Tubing, high temperature for circulators
PTFE lined, stainless steel braid
Flexible PTFE lined, stainless steel over braided tubing with 6,3 mm NPT quick connectors at both ends.
Maximum temperature is 300 °C.
Length (mm): 930
1...,230,231,232,233,234,235,236,237,238,239 241,242,243,244,245,246,247,248,249,250,...612
Powered by FlippingBook