Sargent Welch 2015-2016 - page 16
LABWARE & SUPPLIES
16
Balances
sargentwelch.com
Ohaus
®
AdventurerPro
AnalyticalBalance
TheMost Complete Balance In its Class
Versatility and user-friendly features set this balance apart from the
rest. It counts, sums, holds, and delivers traceable results. With 17
different weighing units, percent weighing, dynamic weighing, a weigh
below hook, and time/date stamping, this balance offers a large num-
ber of features in a compact design. A frameless draft shield allows for
easy access and its design provides superior stability. Removable front
and back glass panels along with a hinged topmake for easy cleaning.
Includes SmarText™ software for easy instruction on each application.
RS232 is built in, options include a second RS232 or USB interface.
Includes two-year warranty.
Cat. No.
Capacity
Model Readability Repeatability PanSize Draft Shield Overall dim.
Linearity
Price
65g x 0.1mg AV64 0.1mg
0.1mg 3.5" dia 9" tall
8.7"W x 12"H x 11.8"D ±0.2g
$2,358.00
110g x 0.1mg AV114 0.1mg
0.1mg 3.5" dia 9" tall
8.7"W x 12"H x 11.8"D ±0.2g
$2,717.00
260g x 0.1mg AV264 0.1mg
0.1mg 3.5" dia 9" tall
8.7"W x 12"H x 11.8"D ±0.3g
$3,229.00
RS232 Interface Cable
$132.00
Cat. No.
Capacity
Model
Readability
Linearity
Platform Size
Price
65
PA64
0.1mg
± 0.2mg
3.5" dia.
$2,138.15
110
PA114
0.1mg
± 0.2mg
3.5" dia.
$2,443.60
210
PA214
0.1mg
± 0.3mg
3.5" dia.
$2,645.00
Ohaus
®
PioneerAnalyticalBalances
Best Balance for BasicWeighing
Designed for routine lab applications, these balances include a draft-
shield with all glass panels and three sliding doors. Each door is
removable for quick and convenient cleaning. An up-front level indicator
eliminates the need to look behind balance during the leveling process.
Three filter modes and adjustable zero tracking are ideal for working
in slow filling applications where sensitivity is required. Three button
operation is user-friendly, and balance features applicationmodes of
weighing, parts counting, and percent. Weighing units aremg, g, oz,
dwt, tical, tola, mommes, baht, grain, mesghl, Newton, ozt, teals, and
custom. Balances include calibration lockout, security loop, weigh below
hook, in use cover, RS232, and an AC adapter. Overall size: 7.7" x
11.3" x 12.6". Includes two-year warranty.
I...,6,7,8,9,10,11,12,13,14,15 17,18,19,20,21,22,23,24,25,26,...795
Powered by FlippingBook