VWR Equip 2016 - page 9
1.800.932.5000 | vwr.com |
equip
9
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
ELECTROCHEMISTRY
HACH
®
H-SeriesBenchtopMeter
–2 to+19pH range
0.1or 0.01pH resolution
7R VQƂXGRQKPV R*ECNKDTCVKQP
#WVQOCVKE QTOCPWCN VGORGTCVWTG EQORGPUCVKQP
6JG* 5GTKGU DGPEJVQROGVGTU CTGFWCN VGEJPQNQI[ R* PQP INCUU +5('6 CPF VTCFKVKQPCN
INCUU 6JGUGRTQHGUUKQPCNOGVGT U[UVGOU UJQYECUG C NCTIG .%&FKURNC[YKVJ .'&DCEMNKIJV CPF
VJG[ JCXG VJG CDKNKV[ VQCEEWTCVGN[OGCUWTGR* +5('6QT INCUU O8 VGORGTCVWTG 142 CPF +5'
6JGUGDGPEJVQROGVGTU CTG GPIKPGGTGF VQIKXG [GCTU QH TGNKCDNG UGTXKEG KP VJGOQUV FKHƂEWNV
of applications.
Description
Includes
Parameters
Cat.No.
Each
Meter,GLPpH,H260G
H260GMeter Only (NoProbe)
Temp, ISFET pH,Glass pH,mV,ORP,Ammonia,Nitrate,
Fluoride, Sodium
1054.38
Meter,GLPpH/COND,H270G
H270GMeter Only (NoProbe)
Temp, ISFET pH,Glass pH,mV,ORP,Ammonia,Nitrate,
Fluoride, Sodium, Conductivity,TDS, Salinity
1308.15
Meter,GLPpH/COND/DO,H280G
H280GMeter Only (NoProbe)
Temp, ISFET pH,Glass pH,mV,ORP,Ammonia,Nitrate,
Fluoride, Sodium, Conductivity,TDS, Salinity,Dissolved
Oxygen
1554.48
HACH
®
TITRALAB
®
AT1000TITRATOR
Automatic TitrationWithout
All theComplications
6JG 6KVTCNCD
®
5GTKGU NCDQTCVQT[ CPCN[UKU U[UVGOU CTGQPG
VQWEJ CWVQOCVKE CPCN[\GTU EQXGTKPIDQVJ2QVGPVKQOGVTKE
6KVTCVKQP #6 5GTKGU CPF-CTN (KUEJGT 8QNWOGVTKE 6KVTCVKQP
-(
5GTKGU YKVJRTG UGVOGVJQFU VJCV ECPSWKEMN[ FGNKXGT
CEEWTCVG CPF TGNKCDNG TGUWNVU UVTCKIJV HTQOVJGDQZ CNNQYKPI
GXGT[QPG KP VJG NCDQTCVQT[ VQ KOOGFKCVGN[ RGTHQTOCEEWTCVG CPF
TGRTQFWEKDNG VKVTCVKQPU
The1000Series features:
Ready-to-use application kits for your unique application
).2 HGCVWTGU KPUKFG HQT TGUWNVU CPF ECNKDTCVKQP VTCEGCDKNKV[
Easy export of data andmethods toUSB (Excel format
EQORCVKDNG
Simple set up
Description
Syringes Pumps
Cat.No.
Each
TitralabAT100Titrator
1
2
3,137.52
TitralabAT100Titrator
2
2
3,541.37
TitralabAT100Titrator
1
-
2,474.35
TitralabAT100Titrator
1
1
2,806.45
Titralab, Karl FisherTitrator
1
2
3,945.22
Visit
vwr.com
formeter bundles.
1,2,3,4,5,6,7,8 10,11,12,13,14,15,16,17,18,19,...24
Powered by FlippingBook