VWR Equip 2016 - page 5
VWR
®
BenchtopChiller, 120V
VWRBenchtopChillers areenvironmentally friendly andeconomical alternatives to tap-water
EQQNKPI 6JGWPKSWGFGUKIPOCZKOK\GU RTGEKQWU DGPEJ URCEGYKVJQWV EQORTQOKUKPIEQQNKPI
andpumpingpower.
(GCVWTGU NCTIG GCU[ VQ TGCF .'&FKURNC[U C URCEG UCXKPIFGUKIP CPF EQQNKPICV CODKGPV
temperatures as high as 35°C.
Compact, space-savingdesign
Integrated safety features
6JG VQR OQWPVGFƂNN RQTVYKVJ URKNN RTQVGEVKQPEWRRTGXGPVU FCOCIG HTQORQVGPVKCN URKNNUYJGP
ƂNNKPI 6JG HTQPV OQWPVGFGCU[ CEEGUU CKT ƂNVGT MGGRU VJG EJKNNGT TWPPKPICV RGCMGHƂEKGPE[
%JQKEGQH RWORU OCIPGVKEFTKXG EGPVTKHWICN VWTDKPG CPFEQORTGUUQT UK\GU
*2
%JKNNGTU CTG UWRRNKGFYKVJ VYQ EO 026 Z EO *$ VWDKPICFCRVGTU VYQ
EO 026 Z EO *$ VWDKPICFCRVGTU CPFRQYGT EQTF 2TKQT VQQTFGTKPI RNGCUG
EJGEM VJG URGEKƂERNWIEQPƂIWTCVKQPPGGFGF HQT [QWT GNGEVTKECN TGSWKTGOGPVU 'WTQRGCPRNWI
QP 8OQFGNU 1VJGT EQWPVT[ URGEKƂERNWIU CTGCNUQCXCKNCDNG
Description
Dimensions
MaximumPressure FlowRate
TemperatureRange
Cat.No.
Each
LSSeriesChillers
1/2HPChiller, Centrifugal Pump
60.7L x 25.4W x 48.3H cm (23.9 x 10 x 19")
14.5psi (1.0bar)
3.5gpm (13.2 L/min.)
–20 to40°C
4278.94
1/2HPChiller, Centrifugal Pump
60.7L x 25.4W x 48.3H cm (23.9 x 10 x 19”)
9.0psi (0.6bar)
3.9gpm (14.8 L/min.)
–20 to40°C
4168.53
1/2HPChiller,Turbine Pump
60.7L x 25.4W x 48.3H cm (23.9 x 10 x 19”)
43.4psi (3.0bar)
2.6gpm (9.8 L/min.)
–20 to 40°C
4599.58
1.800.932.5000 | vwr.com |
equip
5
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
SAMPLE PREP
VWR
®
VacuumOvens
Theseunits featurea front-mounted three-way valve for evacuation, ventingandpurgingof
KPGTV ICUGU GI 0KVTQIGPQT#TIQP YKVJXCEWWOƂVVKPIU QP VJG HTQPV QH VJGWPKV
4CFKCPVYCTOYCNN JGCVKPI U[UVGOQRVKOK\GU WPKHQTOKV[ CPF EQPUGTXGU EJCODGT URCEG HQT
FT[KPI EWTKPI XCEWWOGODGFFKPICPFRNCVKPICRRNKECVKQPU
EO INCUUYQQN KPUWNCVKQPRTGXGPVU JGCV NQUU
Siliconedoor gasket andpositive latchdoormaintain seal at all vacuum levels
2QN[ECTDQPCVG UCHGV[ UJKGNFRTQVGEVU FQQT INCUU
Built-inover-temperatureprotection
Vacuum level is displayedongauges from0 to30 in. Hg
8CEWWOCPFCKT NKPGU CTG EQTTQUKQP TGUKUVCPV UVCKPNGUU UVGGN VWDKPI HQT QRVKOWOEJCODGT
cleanliness and long-termperformance
9KVJ VYQ TGOQXCDNG UVCEMKPICNWOKPWOUJGNXGU
'CU[ VQ ENGCP6[RG UVCKPNGUU UVGGN KPVGTKQT EJCODGT
2QYFGT EQCVGFJGCX[ ICWIG UVGGN GZVGTKQT HQT FWTCDKNKV[
6YQ [GCTYCTTCPV[YKVJGZVGPFGFYCTTCPV[ RCEMCIGU CXCKNCDNG
Volume
ExteriorDimensions
InteriorDimensions
TemperatureRange
Temperature
Uniformity
Fluctuation ShippingWeight
Cat.No.
Each
12.5 L (0.44 cu. ft.)
41× 43×44 cm
(16× 17×17”)
30× 20× 20 cm
(12 x 8 x 8”)
Ambient +10° to 220°C ± 2.2°C at 100°C
25 in.Hg±
±1.5°C
50 kg (110 lbs.)
4215.25
19.8L (0.7 cu. ft.)
40× 50×41 cm
(16× 19.8× 16.25")
30×25×25 cm
(12 x 10 x 10")
Ambient +10° to220°C ±6.0°C at 150°C,
25 in.Hg
±1.0°C
59 kg (130 lbs.)
3760.80
65.1 L (2.3 cu. ft.)
64× 64× 56 cm
(25× 25× 22”)
51×36×36 cm
(20 x 14 x 14”)
Ambient +10° to 220°C ±5.0°C at 100°C,
25 in.Hg
±1.0°C
134 kg (295 lbs.)
7485.02
1,2,3,4 6,7,8,9,10,11,12,13,14,15,...24
Powered by FlippingBook