VWR Equip 2016 - page 4
Description
Cat.No.
Each
OmniMixer Homogenizer
2850.00
OmniMacroHomogenizer
4195.00
Accessories
50mL Stainless Steel Chamberwith1"Blade
995.00
50mL Stainless Steel Chamberwith10mmGenerator Probe
1390.00
400mL Stainless Steel Chamberwith2"Blade
995.00
400mL Stainless Steel Chamberwith20mmGenerator Probe
1725.00
4
equip
| vwr.com | 1.800.932.5000
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
SAMPLE PREP
Since1956 theOmniMixerandMacroHomogenizershavebeensynonymouswith
laboratoryhomogenizers. Referenced inover5,000publications,OmniMixers can
bedependedon for years of trouble-freeoperation.
OmniMixerLaboratoryHomogenizers
OmniMixer
OmniMacro
UnsurpassedQualityandVersatility
600 AND 1800WATT VARIABLE SPEEDMOTORS
COMPATIBLEWITHAWIDEARRAYOFGENERATORPROBES,
SEALEDCHAMBERSANDBLADEASSEMBLIES.
VWR
®
GravityConvectionOven
VWRGravityConvectionovens are theoptimal choice for heating anddryingof delicate
UCORNGU VJCV TGSWKTG CIGPVNG CKTƃQY 2QYFGTU CPF NKIJV UCORNGUYKNN PQV DGDNQYPCYC[
D[ CEVKXG CKTOQXGOGPV 2TQEGUUGU UWEJCU UVGTKNK\CVKQPQT JGCVGF UVQTCIG CTG HQNNQYGF
GEQPQOKECNN[ #KT GZEJCPIG ECPDG EQPVTQNNGFD[ CFCORGT sQRGP KV HQT FT[KPI VQ TGOQXG
humidity quickly, and close it for heat treatmentwithhighest requirements for temperature
CEEWTCE[ %QPUKUVGPE[ s4GRGCVCDNG TGUWNVUYKVJQWVUVCPFKPI VGORGTCVWTG UVCDKNKV[ CPF
uniformity
Versatility –Coverswide rangeof heating anddrying applications
'ZVTGOGN[ XGTUCVKNGYKVJCYKFG VGORGTCVWTG TCPIG Œ%s Œ%CPF VJTGGDGPEJ VQR
sizes ranging from2.3 cu. ft –6.2 cu. ft.
YKTGOGUJ UJGNXGU KPENWFGF NDU ECRCEKV[ GCEJ
6YQ ;GCT9CTTCPV[YKVJ'ZVGPFGF9CTTCPV[ 2CEMCIGU#XCKNCDNG
E%5#WU EGTVKƂGF
Volume
Exterior
Dimensions
PackagingSize
InteriorDimensions Heat-UpTime RecoveryTime
Temperature
Uniformity
Shipping
Weight
Cat.No.
Each
65 L (2.3 cu. ft.)
22D x 21W x 28H" 26D x 30.7W x 39.4H" 16.3D x 13W x 19H" 25min. to 150°C 6min. to150°C ±4°C at 150°C
(basedon27 sensors)
121.25 lbs.
1829.97
105 L (3.7 cu. ft.) 22D x 25W x 32H” 29.9D x 30.7W x 43.7H” 16.3D x 17W x 23H” 24min. to 150°C 8min. to150°C ± 4.5°C at 150°C
(basedon27 sensors)
143.3 lbs.
2180.98
176 L (6.2 cu. ft.) 29D x 25W x 36H” 37.8D x 30.3W x 47.2H” 23D x 17W x 27H” 28min to150°C 8min to 150°C ± 5°C at 150°C
(basedon27 sensors)
189.6 lbs.
2996.97
Specifications
OmniMixerHomogenizer
OmniMacroHomogenizer
Power 115V, 600W
115V, 1800W
VolumeRange 0.25mL to10 L
0.25mL to30 L
SpeedControl
1,000 to18,000 rpm
1,000 to20,000 rpm
1,2,3 5,6,7,8,9,10,11,12,13,14,...24
Powered by FlippingBook