VWR Equip 2016 - page 3
VWR
®
Universal HomogenizingPackage
TheVWR
®
DeluxeUniversal Homogenizing Package is designed for usewith all types of
UCORNGU GXGP VQWIJQT UVTKPI[ 6JKU RCEMCIG CNNQYU VJGƃGZKDKNKV[ VQRTQEGUU VWDGU TCPIKPI
HTQO s O. CUYGNN 6JG OOCPF OOUCY VQQVJIGPGTCVQT RTQDGU RTQXKFG HCUV CPF
GHƂEKGPV QRGTCVKQP (TQOUQHV VQ VQWIJ UCORNGU DQVJ UCY VQQVJIGPGTCVQT RTQDGU RTQXKFG
GZVTC EWVVKPICEVKQPFWTKPIJQOQIGPK\KPI GPUWTKPI HCUV CPF EQORNGVG UCORNGDTGCMFQYP
6JGRCEMCIG KU UWRRNKGF EQORNGVGYKVJ UVCPFCUUGODN[ HQT JCPFU HTGGJQOQIGPK\KPIYKVJKP
UK\GU TCPIKPI HTQOOKETQ VWDGU VQ NCTIG VWDGU YKVJQWV JCXKPI VQJQNF VJGJQOQIGPK\GT WPKV
TheVWR
®
DeluxeUniversal Homogenizing Package includes VWR
®
200Homogenizer
WPKV Z OOUCY VQQVJIGPGTCVQT RTQDG Z OOUCY VQQVJIGPGTCVQT RTQDG CPF
UVCPFCUUGODN[
Description
Cat.No.
Each
VWR®DeluxeUniversal HomogenizingPackage
2988.59
VWR® StandardUniversal HomogenizingPackage
2683.28
VWR
®
Mini HotplateStirrer, 120V
Rugged, compact hotplate stirrers feature an ergonomic design and are ideal for educational
NCDU 6JGUGWPKVU ECPJGCV CPF UVKT WR VQ O. QHYCVGT &WTCDNG Z ECUV CNWOKPWOVQR
platewill not chipor crack, andprovides auniformheating surface. Bi-metallic thermostat
QHHGTU TGNKCDNG VGORGTCVWTG EQPVTQN 2QYGTHWN JGCVGT TGCEJGUOCZKOWOVGORGTCVWTG KP
OKPWVGU 2QYGTHWNOQVQT CPFOCIPGV FGNKXGT TGNKCDNG CPF EQPUKUVGPV UVKTTKPI
6JGDWKNV KP UWRRQTV TQFJQNFGTYKVJ NQEMKPIMPQDCEEGRVU CPQRVKQPCN UWRRQTV TQFCPF
clamp kit. Kit includes 45.7cm (18”) support rod, thermometer extension clamp, three-prong
OGFKWOUYKXGN ENCOR CPFJQQM EQPPGEVQT 1RVKQPCN UWRRQTV RNCVG CNNQYU JQV RNCVG VQDG
OQWPVGF VQ NCD HTCOGU QT TKPI UVCPFU
Ordering Information:
6JG 8WPKVU KPENWFG C EO YKTG EQTFCPFRNWI #NN
WPKVU KPENWFG CƂXG [GCT NKOKVGFYCTTCPV[ QPRCTVU CPF NCDQT
TUV listed. CEmarked. RoHS compliant.
Electrical
MaximumCapacity
SpeedRange
TemperatureRange
Cat.No.
Each
120V, 50/60Hz
1000mL
100–1200 rpm
to 375°C
438.82
1.800.932.5000 | vwr.com |
equip
3
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
SAMPLE PREP
Specifications
Electrical
120V
VolumeRange
1.5-50mLTubes
SpeedRange
Analog: 5,000-35,000 rpm (with SeparateON/OFF Switch)
1,2 4,5,6,7,8,9,10,11,12,13,...24
Powered by FlippingBook