VWR Equip 2016 - page 21
1.800.932.5000 | vwr.com |
equip
21
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
LAB ESSENTIALS
Electrical Volume
Exterior
Dimensions
Door
Type
No. of
Shelves
Temperature
Range
Cat.No.
Each
Refrigerator
115V, 4A
566 L
(20 cu.
ft.)
86.4W x
90.8D x
186H cm
(34 x 35
3
/
4
x 73
1
/
4
")
Single
Solid
Door
with 5
Bins
1
Adjustable,
3Fixed
0 to10°C
(32 to 50°F)
4689.43
Freezer
115V, 4A
566 L
(20 cu.
ft.)
86.4W x
90.8D x
186H cm
(34 x 35
3
/
4
x 73
1
/
4
")
Single
Solid
Door
with 5
Bins
4 Fixed
–18 to–25°C
(–0.4 to
–13°F)
4069.87
VWR
®
Traceable
®
High-Accuracy
Refrigerator
Thermometers
Perfect formonitoring
HTGG\GTU JGCVKPIDNQEMU
KPEWDCVQTU TGHTKIGTCVQTU CPF
YCVGT DCVJU &KURNC[ ECUG KU
madeof high-impact,
EJGOKECN TGUKUVCPV #$5RNCUVKE
Display simultaneously shows
OCZKOWO OKPKOWO CPF EWTTGPV RTQDG VGORGTCVWTGU CUYGNN
CU TQQOVGORGTCVWTG #NCTOHGCVWTGRTQXKFGU EQPVKPWQWU XKUWCN
CPFCWFKQCNGTVUYJGP VGORGTCVWTG TKUGU CDQXGQT HCNNU DGNQY
CRTQITCOOCDNG UGV RQKPV GXGP KH VGORGTCVWTG TGVWTPU VQ
PQP CNCTOTCPIG &WCN RTQDG VJGTOQOGVGTU RTQXKFG
KPFKXKFWCN TGCFKPIU CPFCNGTVU HQT GCEJRTQDG 5GPUQT CPF
O OKETQ ECDNG EQPVKPWG VQRGTHQTOCEEWTCVGN[YJGP
UWDOGTIGF KPYCVGT /KETQ ECDNGRGTOKVU TGHTKIGTCVQT FQQTU VQ
closeon itwithout affecting the seal. Thermometers feature
QPG DWVVQP UGNH ECNKDTCVKQP
6TCEGCDNG
6GORGTCVWTG KP Œ%CPF Œ(
'CEJ UGPUQT RTQDG KU UGCNGF KPCDQVVNGƂNNGFYKVJ)4#5
nontoxic glycol
Description
Range
Accuracy
Cat.No.
Each
CalibratedThermometerwith
NISTTraceableCertificate and
TwoBottle Probes
-58.00 to 158.00°F
(-50.00 to70.00°C) ±0.30°C
115.13
VWR
®
symphony
FlammableStorage
Refrigerator and
Freezer
6Q HCEKNKVCVG VJG UVQTCIGQH ƃCOOCDNG
materials, sources of sparks and ignition –
such as lights, thermostats, switches and
HCPU sJCXGDGGP TGOQXGF HTQOVJG KPVGTKQT
QH VJGUGWPKVU #NN WPKVU JCXG GZVGTKQT
CFLWUVCDNGOGEJCPKECN VJGTOQUVCVU 6JG
GZVGTKQTU HGCVWTG CYJKVGDCMGF QPRQYFGT
EQCV ƂPKUJ CPF UKPING UQNKFFQQTUYKVJMG[GF
locks.
Refrigerators and freezers are suppliedwith
C VYQ [GCT RCTVU CPF NCDQTYCTTCPV[ YKVJCPCFFKVKQPCN VJTGG [GCT
warranty on compressor parts.
'ZVGTKQT CFLWUVCDNGOGEJCPKECN VJGTOQUVCVU
9JKVGDCMGF QPRQYFGT EQCV GZVGTKQT
Single soliddoor,with keyed locks
7PKVUOGGV 0(2#CPF15*#IWKFGNKPGU CPFCTG7. CPF%7. NKUVGF
PROS:
CONS:
Mobile
Noisecanbea factorwith internalblower
Easy installation
Filtermaintenance is required
Energy savings
Limitedapplicationswith limitedfilteroptions
Cheaper installation Greater riskof chemical exposureuponfilter loading
Filter lifeand thusfilter replacement
timesdependon several factors:
• What chemicalsarebeingused, and
filter capacity
• Theevaporation rateofeachchemical
• Thechemical volume that isbeing
used
• Thedurationofusageperday foreach
chemical
• The temperatureofeachchemical
4.Howdo theprosand consof
usingaductless fumehoodbalance
out foryou?
If youhavegotten to thispoint and
havedetermined that aductless fume
hood is suitable for your application,
the last thing todo is todeterminehow
thebenefitsof aductless fumehood
standupagainst itsdisadvantages, and
howdobothalignwithyouroverall
company’sgoals?Thefinal decision
isup toyouandyour safetyofficer.
Whenconsideringwhether topurchasea
DuctlessFumeHood, these four
importantconsiderationswillhelpyou to
make the safest,mosteconomical
decision foryour facility.
1...,11,12,13,14,15,16,17,18,19,20 22,23,24
Powered by FlippingBook