VWR Equip 2016 - page 19
1.800.932.5000 | vwr.com |
equip
19
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
PHYSICALMEASUREMENT
VWR
®
A-SeriesBalances
894# 5GTKGU $CNCPEGU RTQXKFG TGRGCVCDNG CPFRTGEKUGYGKIJKPIYKVJCFXCPEGF
technological features.
State-of-the-art features for advancedweighing anddocumentation
+PVGTPCN ECNKDTCVKQP CWVQOCVKE VKOG QT VGORGTCVWTG CEVKXCVGF
Multipleweighmodes
#FXCPEGF EQOOWPKECVKQP HGCVWTGU
+PCFFKVKQP VQ HQTEGF EQORGPUCVKQPYGKIJKPI VGEJPQNQI[ 894# 5GTKGU
DCNCPEGU KPEQTRQTCVG UVCVG QH VJG CTV HGCVWTGU HQT CFXCPEGFYGKIJKPI
CRRNKECVKQPU CPFFQEWOGPVCVKQP 6JGUG GEQPQOKECN DCNCPEGU QHHGT C
XCTKGV[ QH UVCPFCTF HGCVWTGU UWEJCU KPVGTPCN ECNKDTCVKQP OGPW FTKXGP
operation,multipleweighmodes, advanced communication features,
and varied applicationmodes.
Specifications
Interfaces
RS232 (2),USB-A, andUSB-B
OperatingTemperature
10 to 40°C (50 to 104°F)
Power Requirements
Input: 100–240VAC, 50/60Hz, 0.6A
Capacity
Readability
Repeatability (Std.Dev.)
PanSize
Cat.No.
Each
60/220g
0.01/0.1mg
0.03/0.1mg
85mm (3.35 in)
4963.00
160g
0.1mg
0.1mg
85mm (3.35 in)
2070.49
220g
0.1mg
0.1mg
85mm (3.35 in)
2367.49
310g
0.1mg
0.1mg
85mm (3.35 in)
2590.60
360g
0.001g
0.001g
128 x 128mm (5.04 x 5.04 in)
1731.58
4500g
0.01g
0.01g
195 x 195mm (7.68 x 7.68 in)
1784.73
6000g
0.01g
0.01g
195 x 195mm (7.68 x 7.68 in)
1990.00
VWR
®
B-SeriesBalances
894$ 5GTKGU $CNCPEGU EQODKPG CFXCPEGFYGKIJKPIYKVJ KPVWKVKXGQRGTCVKQP
4GNKCDNG TGUWNVU
Multipleweighmodes
9QTT[ HTGGƂXG [GCTYCTTCPV[
(QTEGF EQORGPUCVKQPYGKIJKPI VGEJPQNQI[
894$ 5GTKGU DCNCPEGU KPEQTRQTCVG UVCVG QH VJG CTV
features for advancedweighing applications. These
GEQPQOKECN DCNCPEGU JCXGFKG ECUV CNWOKPWO
housing, stainless steel weighpans anddraft
EJCODGT ƃQQTU VJCVYKVJUVCPF VJG TKIQTU QH
harsh environments.
Capacity
Readability
Repeatability (Std.Dev.)
PanSize
Cat.No.
Each
60g
0.1mg
0.1mg
90mm (3.5 in)
1625.45
120g
0.1mg
0.1mg
90mm (3.5 in)
1845.60
210g
0.1mg
0.1mg
90mm (3.5 in)
2265.00
200g
0.001g
0.001g
90mm (3.5 in)
1249.00
400g
0.001g
0.001g
90mm (3.5 in)
1548.00
500g
0.001g
0.001g
90mm (3.5 in)
1795.00
4000g
0.01g
0.01g
160mm (6.3 in)
1422.00
5000g
0.01g
0.01g
160mm (6.3 in)
1685.22
1...,9,10,11,12,13,14,15,16,17,18 20,21,22,23,24
Powered by FlippingBook