VWR Equip 2016 - page 18
18
equip
| vwr.com | 1.800.932.5000
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
PHYSICALMEASUREMENT
FungilabDigital Rotational Viscometer
PREMIUMSeries
Premium Series is thehighest performancedigital rotational viscometer for viscosity readings.
9JGPOQPKVQTGFD[ (WPIKNCD&CVC$QUU UQHVYCTG XKUEQOGVGTU QHHGT CYKFG CPFWPKSWG TCPIG
of rheological applications.
/CKP HGCVWTGU KPENWFG )TCRJKE FKURNC[ HQT GCU[OGPWCEEGUU CPF TGUWNVU TGCFKPICPF VQQDVCKP
VJGITCRJKE ƃQYEWTXGDGJCXKQT QH VJG UCORNG CV TGCN VKOG +PENWFGU MG[ VQWEJMG[RCF
TGNCVKXG CDUQNWVG CPFMKPGOCVKE XKUEQUKV[ TGCFKPICWVQOCVKECNN[ WPKV EQPXGTV 5+ VQ%)5
#WVQVGUV CPF#WVQTCPIG HWPEVKQPU YQTMKPIOGOQTKGU CPFRTQITCOOKPI HGCVWTGU CU UVGR
OWNVKUVGRCPF TCOR VKOG VQ VQTSWG CPF VKOG VQ UVQR VGORGTCVWTG TGCFKPI26 +PVGTHCEG
USB connection; 10 languageoptions. Datalogger software included for downloadingdata to
computer towork in Excel
®
ƂNG YKVJ75$ EQPPGEVQT
(WPIKNCD&CVC$QUU 5QHVYCTG HQT TJGQNQIKECN UVWFKGU CPF EQPVTQN WPKV VJTQWIJ VJG 2% KU
QRVKQPCN 6JG (WPIKNCD&CVC$QUU UQHVYCTGRTQXKFGU KPUVCPVCPGQWU XKUEQUKV[ ƃQYU EWTXGU 7R
VQ HQWT FCVC UGVUOC[ DGRNQVVGF UKOWNVCPGQWUN[ /CVJOQFGNKPI HQT [KGNF UVTGUU ECNEWNCVKQPU
$KPIJCO2NCUVKE %CUUQP 2QYGT .CY +2%2CUVG CPCN[UKU /QTG VJCP XKUEQUKV[ RNQVU ECPDG
EQPƂIWTGF 1HHGTU XGT[ GCU[ ITCRJKE RTGUGPVCVKQPQH FCVC CPF UVQTGU VJGFCVC KPC/KETQUQHV
#EEGUU FCVCDCUG
VWR
®
Glass TubeFlowMeters,GRSeries
$QTQUKNKECVGINCUU VWDGƃQYOGVGTU KP OO
9
/
16
UECNG NGPIVJU HGCVWTG C TQVCT[
OCIPKƂECVKQP NGPU CPFƃQYEWTXGU VJCV RGTOKVOWNVKRNGƃWKFCPFICU WUGYKVJC UKPING VWDG
)NCUU VWDG CNNQYU EQORCVKDKNKV[YKVJOQUV ICUGU CPFƃWKFU
(NQYOGVGTU KPENWFG (062 CNWOKPWOƂVVKPIU CPF UVCPFCTF KPNGV XCNXG #NWOKPWOƂVVKPIU
KPENWFG$WPC 0UGCNU
MeasurementRange
Cat.No.
Each
200 to 106.000.000 cP.
5210.23
AirFlowrate
FloatMaterial
WaterFlowrate
Cat.No.
Each
0.24–75.8CCM
Sapphire
184.17
0.405–49.9CCM
BlackGlass
184.17
1.87–278CCM
Carboloy
184.17
238–11,500CCM
Stainless Steel
4.16–336CCM
184.17
3.34–311CCM
Tantalum
205.90
3070–65,800CCM
Tantalum
80–2058CCM
219.26
47.8–857CCM
Stainless Steel
184.17
63–1804CCM
Stainless Steel
1.06–50.2CCM
184.17
1...,8,9,10,11,12,13,14,15,16,17 19,20,21,22,23,24
Powered by FlippingBook