VWR Equip 2016 - page 16
SPECTROSCOPY
QuickLock forDraftShields
Dismantle to clean
in seconds
7" Touchscreen Interface
Makesyour
smartphone look bad
SophisticatedSecurity
Goodbyeweighing
errors
Model
Capacity andReadability
VWRCatalogNo.
Each
MS104TS
120g x 0.1mg
4907.70
MS204TS
220g x 0.1mg
5940.90
MS303TS
320g x 1mg
3337.24
MS1602TS 1620g x 0.01 g
3016.94
MS3002TS 3200g x 0.01 g
3533.54
*Additional models available on us.vwr.com
Combining70 years of weighing expertisewith brilliant intuition, designed
for the everyday user.
Nomorewaiting, nomore downtime. To ensure immediate delivery, the
bestsellingmodels are now guaranteed in-stock!
Call
(800) 932-5000
or visit
us.vwr.com
to learnmore.
SophisticationMadeSimple
TheNewMS-TSBalance
$200
Receive aCashRebate
on your next purchase!
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
PHYSICALMEASUREMENT
1...,6,7,8,9,10,11,12,13,14,15 17,18,19,20,21,22,23,24
Powered by FlippingBook