VWR Equip 2016 - page 15
VWRoffersawide rangeof equipment and instrumentation tomeet your
needsandenable science inanyworkflow.
Inaddition toexcellentproduct choice, VWRprovides technical services
to support you inall phasesof your equipment and instrumentation
applications. VWRoffers technical support includingcalibration,
installation,warranty, and leasing services.
Visit
vwr.com/equip-instruments
formore information.
EQUIPMENTAND INSTRUMENTATION
1.800.932.5000 | vwr.com |
equip
15
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
SPECTROSCOPY
HowMuchSampleDo YouNeed?
WithUV/VIS Excellence–OneDrop
The UV5Nano Spectrophotometer fromMETTLER TOLEDO only
requires 1μL of sample for reliablemeasurements. Patented
LockPath
6GEJPQNQI[ CWVQOCVKECNN[OGCUWTGU CV VYQ FGƂPGF
path lengths, allowing awide concentration range to be
measuredwithout further dilutions. Minimize your sample,
maximize performance.
Call
(800) 932-5000
or visit
vwr.com
to learnmore.
Model
Mettler
Part No.
VWR
Cat. No.
Each
UV5 Excellence
30254725
9,000.00
UV5Bio Excellence
30254728
9,800.00
UV5Nano Excellence
30254729
12,000.00
1...,5,6,7,8,9,10,11,12,13,14 16,17,18,19,20,21,22,23,24
Powered by FlippingBook