VWR Equip 2016 - page 14
14
equip
| vwr.com | 1.800.932.5000
Formore informationon specificationsand
accessories, simplyenter theCat.No. on
vwr.com.
SPECTROSCOPY
B&WTekNanoRamHandheldRamanSystem
6JG0CPQ4COKU C UVCVG QH VJG CTV JCPFJGNF4COCP KPUVTWOGPV HQT PQPFGUVTWEVKXG
KFGPVKƂECVKQPCPF XGTKƂECVKQPQHOCVGTKCNU UWEJCU#2+U GZEKRKGPVU KPVGTOGFKCVGU CPFƂPKUJGF
RTQFWEVU %QORCEV CPFCIKNG VJG0CPQ4COECPDGWUGFD[ PQP VGEJPKECN WUGTU VQ TCRKFN[
KFGPVKH[ UCORNGU KP VJG NCD YCTGJQWUG NQCFKPIFQEMQT ƂGNF JGNRKPI VQGNKOKPCVGSWCTCPVKPG
areas and expeditematerials through themanufacturing lifecycle. UtilizingRaman technology,
PQP EQPVCEV CPCN[UKU ECPDGRGTHQTOGF VJTQWIJ VTCPURCTGPV EQPVCKPGTU CNN YJKNGOCKPVCKPKPI
the volume and integrity of the sample.
*KIJ DTKIJVPGUU VQWEJ UETGGPFKURNC[
'ODGFFGF
FKOGPUKQPCN DCTEQFG UECPPGT
IP-64dust tight and splashproof ratedhousing
Sampling accessories for almost any environment
Batch scanningoption for large volumeoperation
+PVWKVKXG UQHVYCTG HQT VGEJPKECN
PQP VGEJPKECN WUGTU
9K (K
GVJGTPGV EQOOWPKECVKQP HQT FCVC U[PE OCPCIGOGPV
ShimadzuBioSpec-nanoSmall-volumeUV
Spectrophotometer
#UVCVG QH VJG CTV788+55RGEVTQRJQVQOGVGT HQT NKHG UEKGPEGU VJG UOCNN XQNWOG EQORNGVGN[
automatedBioSpec-nano features anoutstandinganalysis rangeandexcellentmeasurement
TGRTQFWEKDKNKV[ OCMKPI KV VJG UQNWVKQP HQT UVTGUU HTGGPWENGKE CEKFSWCPVKVCVKQPCPFRTQVGKPCPCN[UKU
-G[ HGCVWTGU KPENWFGWNVTC UOCNN UCORNG XQNWOGU Ş. Ş. CWVQOCVKE UCORNGOQWPVKPI
OGCUWTGOGPV CPF ENGCPKPI CPFGCU[ QRGTCVKQPYKVJ &TQR CPF %NKEM CPCN[UKU +PCFFKVKQP VJG
$KQ5RGEPCPQGPUWTGU GZEGNNGPVOGCUWTGOGPV TGRTQFWEKDKNKV[ GPCDNGU RJQVQOGVTKE CPCN[UKU WR
VQ YCXGNGPIVJU CPF ECPDG GSWKRRGFYKVJCPQRVKQPCN OORCVJNGPIVJ EGNN
VWR
®
Spectrophotometers
Fully scanning spectrophotometer foradvancedmeasurementsandqualitycontrol.Reliable, robustand
easy touse instrumentswitha rangeofaccessories thatwill support thecreationofapersonal
spectrophotometermeasuring station. Theapplication softwareoffersall the importantmethods
expected froma ‘universal’ instrument.Allhoused ina robustcoverwithawelldesignedopticalbench.
Description
Cat.No.
Each
NanoRamHandheldRaman Spectrometer Package
35,558.78
Description
Cat.No.
Each
BioSpec-nano Small-volumeUV Spectrophotometer
9821.90
Description
Bandwidth
Calibration
LightType
Wavelength
Weight
Cat.No.
Case of 1
V-3000PC,VIS Scanning Spectrophotometer
4
±0,002A
(320 - 1000nm)
TungstenHalogen,±0,05%T
at 360nm
320 - 1100nm 12 kg
3990.55
UV-3100PC,UV/VIS Scanning Spectrophotometer 2
±0,002A
(200 - 1000nm)
Deuterium/TungstenHalogen, Stray
Light ±0,05%T@220, 360nm
| |
PO 14 kg
5012.71
1...,4,5,6,7,8,9,10,11,12,13 15,16,17,18,19,20,21,22,23,...24
Powered by FlippingBook