Ward's Science Physics & Chemistry - page 218
218
chemicals
|
chemistry
Oil ofWintergreen
SeeMethylSalicylate
OleicAcid
CH
3
(CH
2
)
7
CH:CH(CH
2
)
7
COOH
F.W.: 282.47
CAS#: 112-80-1
Shelf Life (months):
12
Storage:
Green
Soluble:
Organic Solvents (Not Water)
General
Storage
bp ( °C):
360
mp ( °C):
13.4
Density (g/mL):
0.89
Grade: Laboratory
9463106
500mL Bottle
OliveOil
CAS#: 8001-25-0
Shelf Life (months):
36
Storage:
Green
Soluble:
Carbon Disulfide, Chloroform
General
Storage
and Ether
Density (g/mL):
0.913
Grade: Laboratory
9463206
500mL Bottle
Orange IV
TropaeolinOO,AcidOrange5
Indicator, transition range
1.4 (red)–2.6 (yellow).
C
18
H
14
N
3
NaO
3
S
F.W.: 375.38
CAS#: 554-73-4
Shelf Life (months):
36
Storage:
Green
Soluble:
Water and Alcohol
General
Storage
Grade: Laboratory, Powder
9463302
25 g Bottle
OxalicAcidDihydrate
HO
2
CCO
2
H • 2H
2
O
F.W.: 126.07
CAS#: 6153-56-6
Shelf Life (months):
36
Storage:
White
Soluble:
Water and Organic Solvents
CORROSIVE
mp ( °C):
101.5
Density (g/mL):
1.65
Grade: Laboratory, Crystal
9717506
500 g Bottle
Pancreatin (cont.)
Solution, 2% (Aqueous)
9463704
100mL Bottle
9463706
500mL Bottle
Papain
Proteolytic enzyme for tissuemaceration.
CAS#: 9001-73-4
Shelf Life (months):
36
Storage:
Green
Soluble:
Water and Glycerol
General
Storage
Grade: Laboratory, Powder
9463804
100 g Bottle
Para nOil
SeeMineralOil
Para nWax
CAS#: 8002-74-2
Shelf Life (months):
36
Storage:
Green
Soluble:
WarmAlcohol, Chloroform,
General
Storage
Petroleum Ether, Olive Oil and Other
Organic Solvents
bp ( °C):
322
mp ( °C):
58–62
Grade: Laboratory
9464006
500 g Bottle
Para nWax,
Re nedWhite
CAS#: 8002-74-2
Shelf Life (months):
36
Storage:
Green
Soluble:
WarmAlcohol, Chloroform,
General
Storage
Petroleum Ether, Olive Oil, and Other
Organic Solvents
bp ( °C):
322
mp ( °C):
58–62
Grade: Laboratory
9463906
500 g Bottle
PBS
SeePhosphateBu eredSaline
Pentane
C
5
H
12
F.W.: 72.15
CAS#: 109-66-0
Shelf Life (months):
36
Storage:
Red
bp ( °C):
36
Density (g/mL):
0.62
Grade: Reagent
9511004
100mL Bottle
Grade: Laboratory
9511107
1 L Bottle
PeppermintOil
CAS#: 8006-90-4
Shelf Life (months):
36
Storage:
Green
Soluble:
Water and Alcohol
General
Storage
Grade: Laboratory
9464402
25mL Bottle
Pepsin
CAS#: 9001-75-6
Shelf Life (months):
12
Storage:
Green
Soluble:
Water
General
Storage
Grade: Laboratory, Powder 1:3,000
9464602
25 g Bottle
9464604
100 g Bottle
Grade: Laboratory, Powder 1:10,000
9464502
25 g Bottle
Peptone
For bacteriological use.
Shelf Life (months):
12
Storage:
Green
General
Storage
Grade: Laboratory, Media
9464704
100 g Bottle
Petrolatum(White)
PetroleumJelly
CAS#: 8009-03-8
Shelf Life (months):
36
Storage:
Green
Soluble:
Organic Solvents (Not Water)
General
Storage
mp ( °C):
38–45
Density (g/mL):
0.85
Grade: Laboratory, Jelly
9464806
500 g Bottle
PetroleumEther
Ligroin
CAS#: 8032-32-4
Hazard:
Flammable
Shelf Life (months):
12
Storage:
Red
Soluble:
Miscible with Organic Solvents
bp ( °C):
35–60
Grade: Reagent
9511204
100mL Bottle
9511206
500mL Bottle
9511207
1 L Bottle
9511208
4 L Bottle
Pancreatin
CAS#: 8049-47-6
Shelf Life (months):
36
Storage:
Green
Soluble:
Partially, inWater
General
Storage
Grade: Laboratory, Powder
9463502
25 g Bottle
9463504
100 g Bottle
1,10-Phenanthroline
Monohydrate
o-Phenanthroline
C
12
H
8
N
2
• H
2
0
F.W.: 128.22
CAS#: 5144-89-8
1...,208,209,210,211,212,213,214,215,216,217 219,220,221,222,223,224,225,226,227,228,...299
Powered by FlippingBook