VWR Collection Laboratory Equipment - page 99
97
VWR
®
CLAMPS
VWR
®
Lab-Frame Kits
VWR
®
offers the widest choice of standard frames available, plus the accessories to custom-fit any laboratory with the right frame for the job. Standard
Lab-Frame sets are available in five sizes. Options of traditional center-less ground aluminum, non-corroding fiberglass, or tough stainless steel rods are
available. Nickel-plated zinc connectors and lab-frame feet are standard with all kits.
VWR
®
Medium Lab-Frame
Horizontal or vertical mounting, convenient for distillation or general set-up.
Frame measures 24 x 48” (610 x 1219mm). Base is 18” (457mm) wide for
stability.
Components: (8) 2” (51mm) rods
(2) 18” (457mm) rods
(7) 24” (610mm) rods
(4) 48” (1219mm) rods
(35) S-Connectors
(4) Rod End Connectors
(4) Lab-Frame Feet
VWR
®
Small Lab-Frame
Vertical mounting, great for glassware set-ups in small laboratories or where
space is limited. Frame measures 24 x 24” (610 x 610mm). Base is 18” (457mm)
wide for stability. Base may be permanently mounted to bench top with screws
(included).
Components: (8) 2” (51mm) rods
(2) 18” (457mm) rods
(8) 24” (610mm) rods
(18) S-Connectors
(4) Rod End Connectors
(4) Lab-Frame Feet
VWR
®
Talon
®
Lab-Frames
Frame Dimensions
Base Dimensions
Aluminum
Cat. No.
Fiberglass
Cat. No.
Stainless Steel
Cat. No.
24 x 24” (610 x 610mm)
18” (457mm) wide
Frame Dimensions
Base Dimensions
Aluminum
Cat. No.
Fiberglass
Cat. No.
Stainless Steel
Cat. No.
24 x 48” (610 x 1219mm)
18” (457mm) wide
1...,89,90,91,92,93,94,95,96,97,98 100,101,102,103,104,105,106,107,108,109,...116
Powered by FlippingBook