VWR Collection Laboratory Equipment - page 98
96
VWR
®
Frame Rods
Everything you need to customize a lab-frame to your laboratory space. Quality construction
throughout. Frame rods are available in many lengths and materials.
Aluminum
Center-less ground for precise fit. These hard aluminum rods provide a smooth fit with
lab-frames. Corrosion resistant with chamfered ends for easy location.
Fiberglass
Strong and resilient, fiberglass rods will not corrode or rust. Structurally strong and resilient
to clamp compression scars.
Stainless Steel
Heavy-duty, top-of-the-line rods provide maximum strength and durability. Constructed of
303 stainless steel.
VWR
®
Lab-Frame Foot
Stainless steel electro-polished finish or nickel-plated zinc construction. Strong, durable lab-
frame foot is designed for mounting lab-frames permanently to bench tops, fume hoods, walls,
and floors. Spilled fluids roll right off the smooth body contour, minimizing contamination and
easing clean-ups. Features a set screw and includes three 0.75” (19mm) mounting screws.
Material
Min. to Max. Grip Size
Dimensions (Dia. x H)
Cat. No.
Stainless Steel
0 to 0.51” (0 to 13mm)
2.75 x 1.25” (70 x 32mm )
Nickel-Plated Zinc
0 to 0.51” (0 to 13mm)
2.75 x 1.25” (70 x 32mm )
aluminum
stainless steel
fiberglass
Aluminum
Fiberglass
Stainless Steel
Rod Diameter
Rod Length
Cat. No.
Cat. No.
Cat. No.
0.51” (13mm)
1.61” (41mm)
0.51” (13mm)
2” (51mm)
0.51” (13mm)
6” (152mm)
0.51” (13mm)
12” (305mm)
0.51” (13mm)
18” (457mm)
0.51” (13mm)
24” (610mm)
0.51” (13mm)
36” (914mm)
0.51” (13mm)
48” (1219mm)
0.51” (13mm)
60” (1524mm)
0.51” (13mm)
72” (1829mm)
0.51” (13mm)
96” (2438mm)
(shown in stainless steel)
VWR
®
Talon
®
Lab-Frames
VWR
®
CLAMPS
1...,88,89,90,91,92,93,94,95,96,97 99,100,101,102,103,104,105,106,107,108,...116
Powered by FlippingBook