VWR Collection Laboratory Equipment - page 97
95
VWR
®
CLAMPS
Max. Weight Mounted
Min. to Max. Grip
Cat. No.
2.27kg (5lbs)
0 to 20mm (0 to 0.78”)
VWR
®
Universal Stirrer Mounting Bracket
Use to mount stirrers and other devices to lab-frames. Aluminum mounting
bracket with attachment screws.
Length
Cat. No.
79mm (3.11”)
VWR
®
Wrench
Special adjusting wrench is for use with lab-frame items with set screws.
Bar Length
Cat. No.
51mm (2”)
102mm (4”)
152mm (6”)
203mm (8”)
VWR
®
Horizontal Mounting Bars with Coupler
Threaded end for direct installation into channels. Comes with spring coupler
for use in steel 41mm (1.61”) channels. Locking nut included. Use with steel
channel frames.
Dimensions
Cat. No.
33 x 40mm (1.3 x 1.58”)
VWR
®
Channel Connector
Special connector fastens lab-frame rod to steel 41mm (1.61”) channels. Loosen
the connector and slide up or down for easy adjustment of rod position.
Min. to Max. Rod Size
Cat. No.
0 to 13mm (0 to 0.51”)
VWR
®
Multi-Rod Connector
Maximum adjustment capability within one connector. Two adjustment rods fit
into the oval hole in the connector, preventing turning when tightened. Side and
front 13mm (0.51”) holes allow creation of a variety of configurations. Nickel-plated
zinc construction.
VWR
®
Talon
®
Connectors & Holders
1...,87,88,89,90,91,92,93,94,95,96 98,99,100,101,102,103,104,105,106,107,...116
Powered by FlippingBook