VWR Collection Laboratory Equipment - page 96
94
Min. to Max. Grip Size
Cat. No.
0 to 13mm (0 to 0.51”)
VWR
®
Frame Connector (pack of 12)
Improved contour delivers simplicity and strength. Angled adjustment
screws allow easy set-up and prevent misalignment. Smooth,
rounded surfaces are easy-to-clean. Small size maximizes lab-frame
space. Bright dipped aluminum finish sealed with silicon for maximum
protection against corrosion.
Material
Min. to Max. Grip Size
Cat. No.
Stainless steel
0 to 13mm (0 to 0.51”)
Nickel-plated zinc
0 to 13mm (0 to 0.51”)
VWR
®
S-Connector
Stainless steel electro-polished finish or nickel-plated zinc. Clamp
is ideal for constructing lab-frames or other supports requiring the
connection of two perpendicular rods. Clamp connects two 13mm
(0.51”) rods at a 90° angle and features separate adjustment screws
for each rod location.
Material
Min. to Max. Grip Size
Cat. No.
Stainless steel
0 to 13mm (0 to 0.51”)
Nickel-plated zinc
0 to 13mm (0 to 0.51”)
VWR
®
Hook Connector
Stainless steel electro-polished finish or nickel-plated zinc construction.
Simple, versatile, and easy-to-use. Hook connectors allow one-handed
assembly of two components with one adjustment screw.
(shown in stainless steel)
(shown in nickel-plated zinc)
VWR
®
End-to-End Connector
Extend the length of lab-frame rods. Strong aluminum alloy connector
permits end-to-end joining of rods. Precision boring of connector
ensures perfect alignment of rods. Corrosion-resistant. Comes with
two set screws.
Min. to Max. Grip Size
Cat. No.
0 to 13mm (0 to 0.51”)
VWR
®
Rod End Connector
Holds rods firmly at 90°. Use when semi-permanent installations
are required. Comes with two set screws and is precision bored for
close fit. Stainless steel electro-polished finish or nickel-plated zinc
construction.
Material
Min. to Max. Rod Size
Cat. No.
Stainless Steel
0 to 13mm (0 to 0.51”)
Nickel-plated zinc
0 to 13mm (0 to 0.51”)
(shown in stainless steel)
(shown in nickel plated -zinc)
(shown in stainless steel)
(shown in nickel plated -zinc)
60079-100
VWR
®
Talon
®
Connectors & Holders
VWR
®
CLAMPS
1...,86,87,88,89,90,91,92,93,94,95 97,98,99,100,101,102,103,104,105,106,...116
Powered by FlippingBook