VWR Collection Laboratory Equipment - page 95
93
VWR
®
CLAMPS
Material
Min. to Max. Grip Size
Cat. No.
Stainless steel
0 to 18mm (0 to 0.71”)
Nickel-plated zinc
0 to 18mm (0 to 0.71”)
VWR
®
Regular Holder
Stainless Steel electro-polished finish or nickel-plated zinc construction.
Ideal for holding clamps to lab-frames. Use wherever clamping at 90° is
required.
Material
Min. to Max. Grip Size
Cat. No.
Stainless steel
0 to 19mm (0 to 0.75”)
Nickel-plated zinc
0 to 19mm (0 to 0.75”)
VWR
®
Swivel Holder
Two rod holders with center swivel capacity allow tilting of
clamps at any angle in parallel planes. Outside adjustment
screw allows close proximity between items being held.
Stainless Steel electro-polished finish or nickel-plated
zinc construction.
Min. to Max. Grip Size
Cat. No.
0 to 19mm (0 to 0.75”)
VWR
®
All-Position Clamp Holder
Surpass standard holding capabilities. The all-position clamp holder
permits adjustment at any angle in any plane. Holders are set at 90°
to each other, connected by a 90° connector, allowing 360° rotation.
Nickel-plated zinc construction.
VWR
®
Jumbo Holder
Stainless steel electro-polished finish or aluminum construction. Ideal
for holding clamps to lab-frames or ring stands.
Material
Min. to Max. Grip Size
Cat. No.
Stainless steel
0 to 21mm (0 to 0.83”)
Aluminum
0 to 21mm (0 to 0.83”)
VWR
®
Heavy-Duty Holder
For mounting stirrers and other apparatus. Clamp is constructed of
strong aluminum alloy; fitted with over-sized knobs for very secure
positioning. Rods supported on 102mm (4”) long surface to avoid
vibration and wobble.
Material
Min. to Max. Grip Size
Cat. No.
Aluminum
6 to 24mm (0.24 to 0.95”)
(shown in nickel-plated zinc)
(shown in aluminum)
(shown in stainless steel)
(shown in stainless steel)
(shown in nickel plated-zinc)
(shown in stainless steel)
VWR
®
Talon
®
Connectors & Holders
1...,85,86,87,88,89,90,91,92,93,94 96,97,98,99,100,101,102,103,104,105,...116
Powered by FlippingBook