VWR Collection Laboratory Equipment - page 94
92
VWR
®
Thumbscrew Knob
Thumbscrew tightening knob features a 32mm (1.25”) slot in the center of a nylon disc that
will accept thumbscrews supplied with the LabJaws and UltraJaws Multi-Purpose Clamps,
excluding the stainless steel version. Makes tightening of clamps easier on the hands and
wrists.
VWR
®
Replacement Sleeves
Sleeves are easily removed for cleaning or replacement. Both vinyl and fiberglass sleeves are available. Fiberglass sleeves are recommended
for all applications above 100°C (212°F).
Overall Diameter
Thumbscrew Slot
Cat. No.
64mm (2.5”)
32mm (1.25”)
Green Vinyl Sleeve
Fiberglass Sleeve
Clamp Size
Cat. No.
Cat. No.
Medium 2-Prong (2 pack)
Large 2-Prong (2 pack)
Small 3-Prong (3 pack)
Medium 3-Prong (3 pack)
Large 3-Prong (3 pack)
VWR
®
Talon
®
Specialty Clamps & Clamp Accessories
VWR
®
CLAMPS
VWR
®
Bench Clamp
Aluminum bracket with arm fastens quickly and firmly to any convenient shelf. Accepts
0.51” (13mm) rods vertically and horizontally, to which ordinary rings and clamps attach,
leaving bench surface clear for apparatus.
Clamp Opening
Depth of Grip
Overall Length
Cat. No.
1.7” (43mm)
2” (51mm)
7.75” (197mm)
1...,84,85,86,87,88,89,90,91,92,93 95,96,97,98,99,100,101,102,103,104,...116
Powered by FlippingBook