VWR Collection Laboratory Equipment - page 93
91
VWR
®
CLAMPS
VWR
®
Ultra Flex Support System with Base Plate
• Stable base plate
• All metal construction
VWR
®
Ultra Flex Support System with Base Plate is designed with a stable, painted steel base
plate with chemical resistant black finish, that fits easily on bench tops or in fume hoods. Base plate
measures 127 x 127 x 12.7mm (5 x 5 x 0.5”).
VWR
®
Ultra Flex Support System with Bench Clamp
• Aluminum bench clamp
• Easily attaches to most lab benches
VWR
®
Ultra Flex Support System with Bench Clamp easily attaches to the side of your lab bench
or counter top, which helps utilize more space in your lab. Bench clamp is aluminum.
VWR
®
Ultra Flex Support System with Lab-Frame Connector
• Cast alloy lab-frame connector
• Mounts to lab-frames or support stands
VWR
®
Ultra Flex Support System with Lab-Frame Connector securely attaches to support stands,
lab-frames, or any support rod up to 19mm (0.75”) in diameter. Ideal for use in fume hoods. Lab-
frame connector is made of cast alloy.
Description
Cat. No.
Ultra Flex 12 with Base Plate
Ultra Flex 18 with Base Plate
VWR
®
Ultra Flex Support Systems have unique flexible arms that are extremely versatile and can be placed in virtually any position or angle. The Ultra Flex
comes in three different systems; base plate, lab-frame connector, and bench clamp. Each system includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring
Clamp Head, nickel-plated flex arm in 305mm (12”) or 457mm (18”) length, and utility wrench. 2-prong and 3-prong clamp heads also supplied with non-slip vinyl
sleeves, and for temperatures above 100°C, fiberglass covers.
Arm Length,
Ultra Flex 12:
305mm (12”)
Ultra Flex 18:
457mm (18”)
Arm Diameter:
13mm (0.51”)
Max. Grip Size,
2-Prong Clamp Head:
75mm (2.95”)
3-Prong Clamp Head:
69mm (2.72”)
Spring Clamp Head:
13mm (0.51”)
2-Prong Clamp Head
3-Prong Clamp Head
Spring Clamp Head
Description
Cat. No.
Ultra Flex 12 with Lab-Frame Connector
Ultra Flex 18 with Lab-Frame Connector
Description
Cat. No.
Ultra Flex 12 with Bench Clamp
Ultra Flex 18 with Bench Clamp
Description
Cat. No.
2- Prong Clamp Head
3- Prong Clamp Head
Spring Clamp Head
12” Flex Arm
18” Flex Arm
Base Plate
Lab-Frame Connector
Bench Clamp
Replacement Parts
VWR
®
Talon
®
Specialty Clamps
1...,83,84,85,86,87,88,89,90,91,92 94,95,96,97,98,99,100,101,102,103,...116
Powered by FlippingBook