VWR Collection Laboratory Equipment - page 92
90
Flask Size
Inside Dia.
Cat. No.
500mL, 1000mL
125mm (4.92”)
2000mL, 3000mL, 4000mL
142mm (5.59”)
VWR
®
Kettle Clamps
Holds flask and cover together firmly. Three insulated spring-activated clamping arms. Stainless steel springs.
For use on reaction kettles.
VWR
®
Buret Holders
Stainless steel electro-polished finish or nickel-plated zinc Double Buret Clamp holds any size buret from
micro to 100mL capacity. Simply compress the scissor-like mechanism, insert buret and gently release to grip.
Numbers and graduation on buret remain easy to read. For height adjustments, recompress mechanism and
slide buret up or down and gently release. Clamp unit attaches to optional standard support rod with built-in
reinforced hook connector. Adjusting nut faces forward for easy use. Stainless steel or aluminum support rod
attaches to optional porcelain base.
Support Rod (dia. x L):
13 x 578mm (0.51 x 22.76”)
Porcelain Base (L x W x H): 178 x 330 x 25mm (7 x 13 x 1”)
Overall Length
Prong Width
Cat. No.
152mm (5.99”)
25mm (1”)
VWR
®
Support Clamp
Supports electrolysis apparatus, glass rods or tubing at any angle. Arms are nickel-plated brass with vinyl
coating. Easily supports cylindrical objects up to 19mm (0.75”) diameter. Built-in holder grips rods up to 19mm
(0.75”) in diameter.
Overall Length
Cat. No.
178mm (7”)
VWR
®
Electrode Support Clamp
Permits suspension of electrodes over beaker for potentiometric titrations. Holds any electrode clamp at
desired level. Built-in nickel-plated zinc holder grips rods up to 19mm (0.75”) in diameter.
Overall Length
Cat. No.
137mm (5.39”)
VWR
®
Suspension Clamp
Holds thermometers, potash bulbs or drying tubes 114mm (4.49”) from support rod. Machine tapered, nickel-
plated brass hook won’t bend, rust, or corrode. Built-in holder grips rods up to 19mm (0.75”) in diameter.
Description
Cat. No.
Double Buret Clamp (only)
Support Stand with Rod
Double Buret Clamp & Support Stand with Rod (complete)
Aluminum
Description
Cat. No.
Double Buret Clamp (only)
Support Rod (only)
Support stand with Rod
Double Buret Clamp & Support Stand with Rod (complete)
Stainless Steel
flask not
included
(shown in
stainless steel)
Electrode holder not included
VWR
®
Talon
®
Specialty Clamps
VWR
®
CLAMPS
1...,82,83,84,85,86,87,88,89,90,91 93,94,95,96,97,98,99,100,101,102,...116
Powered by FlippingBook