VWR Collection Laboratory Equipment - page 90
88
Min. to Max. Grip Size
Overall Length
Cat. No.
6 to 8mm (0.24 to 0.32”)
159mm (6.26”)
VWR
®
Thermometer Swivel Clamp
Holds glass tubes and thermometers 114mm (4.49”) from support rod. Clamp features safety
adjust spring plate jaws that adjust to any angle with locking wing nut. Built-in holder grips
rods up to 19mm (0.75”) in diameter. Lightweight, rust, and corrosion-resistant. Nickel-plated
construction.
Min. to Max. Grip Size Ext Arm Length Arm Dia.
Overall Length
Cat. No.
6 to 12mm (0.24 to 0.47”)
127mm (5”)
11mm (0.43”)
210mm (8.27”)
VWR
®
Thermometer / Thermocouple Extension Clamp
Lightweight clamp holds glass tubing/thermometers or thermocouples up to 178mm (7”) from
support rod. Tightening wing nut applies tension to the nickel-plated jaws.
NOTE
: An additional holder must be purchased in order to attach clamps to frames or other
apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
Min. to Max. Grip Size
Overall Length
Cat. No.
5 to 10mm (0.2 to 0.39”)
80mm (3.15”)
VWR
®
Wall Clamp
Ideal for securing fermentation tubes, burets or other small objects to walls where no frames are
available. Integral self-tapping screw secures clamp to wall. Nickel-plated construction.
Size
Min. to Max. Grip Size
Prong Width Overall Length
Cat. No.
Small
5 to 10mm (0.2 to 0.39”)
N/A
89mm (3.5”)
Large
0 to 46mm (0 to 1.81”)
11mm (0.43”)
117mm (4.61”)
VWR
®
Water Bath Clamps
Holds a variety of apparatus, including glass tubes and thermometers onto glass water bath walls.
Built-in holder grips walls of varying thickness, up to 9mm (0.35”). Knurled thumbscrew tightens
jaws to hold objects firmly. Nickel-plated zinc construction. Large water bath clamp is supplied
with non-slip vinyl sleeves, and for temperatures above 100°C, fiberglass covers.
NOTE
: Replacement vinyl and fiberglass sleeves are available, page 92.
VWR
®
Talon
®
Specialty Clamps
VWR
®
CLAMPS
1...,80,81,82,83,84,85,86,87,88,89 91,92,93,94,95,96,97,98,99,100,...116
Powered by FlippingBook