VWR Collection Laboratory Equipment - page 9
7
VWR
®
SHAKERS
* Maximum speed/tilt angle may vary with heavy or unbalanced loads
** Centered on tray.
Orbital Shaker
Advanced 5000
Orbital Shaker
Advanced 10000-1
Orbital Shaker
Advanced 10000-2
Orbital Shaker
Advanced 15000-1
Orbital Shaker
Advanced 15000-2
20 to 500rpm
15 to 500rpm
15 to 500rpm
15 to 500rpm
15 to 500rpm
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
Orbital
Orbital
Orbital
Orbital
Orbital
25mm
25mm
51mm
25mm
51mm
50lbs
100lbs
100lbs
150lbs
150lbs
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
YES
18 x 24”
24 x 24”
24 x 24”
24 x 36”
24 x 36”
18 x 24”, 18 x 30”
Adjustable Platform
Large Vessel Carrier Platform
24 x 24”
Large Vessel Carrier Platform
24 x 24”
Large Vessel Carrier Platform
24 x 36”
Large Vessel Carrier Platform
24 x 36”
Large Vessel Carrier Platform
High Speed Microplate Shaker
Advanced
Rocking Platform
Shaker
3-D Rotator
Waver
600 to 2500rpm
1 to 50rpm*
1 to 30rpm*
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
Orbital
Rocking
Waving
3.6mm
Tilt Angle: 0 to 15° *
Tilt Angle: 0 to 20° *
48 microplates
10lbs**
5lbs**
NO
YES
YES
NO
NO
NO
DC Motor
Stepper Motor
Stepper Motor
NO
YES
YES
NO
YES
YES
NO
NO
NO
YES
NO
NO
11 x 12.25”
12.75 x 10”
11.75 x 8.75”
N/A
Stacking Tray
Stacking Tray
VWR
®
Open-Air Shaker Selection Guide
1,2,3,4,5,6,7,8 10,11,12,13,14,15,16,17,18,19,...116
Powered by FlippingBook