VWR Collection Laboratory Equipment - page 89
87
VWR
®
CLAMPS
Material
Description
Size
Min. to Max. Grip Size
Prong Width
Overall Length
Cat. No.
Stainless steel
2-Prong Single Adjust
Medium
0 to 77mm (0 to 3.03”)
23mm (0.91”)
133mm (5.24”)
Nickel-plated zinc
2-Prong Single Adjust
Medium
0 to 77mm (0 to 3.03”)
23mm (0.91”)
133mm (5.24”)
Stainless steel
3-Prong Dual Adjust
Medium
0 to 69mm (0 to 2.72”)
20mm (0.79”)
146mm (5.75”)
Nickel-plated zinc
3-Prong Dual Adjust
Medium
0 to 69mm (0 to 2.72”)
20mm (0.79”)
146mm (5.75”)
VWR
®
Swivel Clamps
Used to hold apparatus near the lab-frame. Unlike extension clamps, the swivel
clamps have an integral holder for attaching to a lab-frame or other apparatus.
Built-in holder grips rods up to 19mm (0.75”) in diameter and is adjustable for
forward or reverse-facing adjustment screws. Shaft wing-nut allows the holding
angle of the swivel clamp to adjust through 360° of rotation and can be locked
in place once desired position is achieved. Stainless steel electro-polished
finish or nickel-plated construction. Supplied with non-slip vinyl sleeves, and for
temperatures above 100°C, fiberglass covers.
NOTE
: Replacement vinyl and fiberglass sleeves are available, page 92.
VWR
®
Fixed-Position Clamps
Used to hold apparatus near the lab-frame where no adjustment is required after
set-up. Built-in holder grips rods up to 19mm (0.75”) in diameter. Fixed-position
clamps have an integral holder but can be rotated after attachment to a lab-frame
or other apparatus. Available in 2-prong or 3-prong. Stainless steel electro-
polished finish or nickel-plated zinc construction. Supplied with non-slip vinyl
sleeves, and for temperatures above 100°C, fiberglass covers.
NOTE
: Replacement vinyl and fiberglass sleeves are available, page 92.
VWR
®
Double Jaw Utility Clamp
Swivels 360° and locks securely in desired position. These 2-prong clamps have a medium and
a large clamp on each end. Nickel-plated zinc construction. Supplied with non-slip vinyl sleeves,
and for temperatures above 100°C, fiberglass covers.
NOTE
: Replacement vinyl and fiberglass sleeves are available, page 92.
Material
Description
Size
Min. to Max. Grip Size
Prong Width
Overall Length
Cat. No.
Stainless steel
2-Prong Single Adjust
Medium
0 to 76mm (0 to 3”)
23mm (0.91”)
163mm (6.42”)
Nickel-plated zinc
2-Prong Single Adjust
Medium
0 to 76mm (0 to 3”)
23mm (0.91”)
163mm (6.42”)
Nickel-plated zinc
2-Prong Single Adjust
Large
0 to 95mm (0 to 3.74”)
23mm (0.91”)
180mm (7.09”)
Stainless steel
3-Prong Dual Adjust
Medium
0 to 69mm (0 to 2.72”)
20mm (0.79”)
178mm (7.01”)
Nickel-plated zinc
3-Prong Dual Adjust
Medium
0 to 69mm (0 to 2.72”)
20mm (0.79”)
178mm (7.01”)
(shown in stainless steel)
(shown in nickel-plated zinc)
(shown in stainless steel)
(shown in nickel-plated zinc)
VWR
®
Talon
®
Specialty Clamps
Description
Min. to Max. Grip Size Prong Width Overall Length
Cat. No.
Medium 2-Prong 0 to 76mm (0 to 3”)
23mm (0.91”) 222mm (8.74”)
Large 2-Prong
0 to 95mm (0 to 3.74”)
1...,79,80,81,82,83,84,85,86,87,88 90,91,92,93,94,95,96,97,98,99,...116
Powered by FlippingBook