VWR Collection Laboratory Equipment - page 88
86
Material
Size (A) Min. to Max. Grip Size (B) Prong Width (C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Nickel-plated zinc Medium 0 to 78mm (0 to 3.07”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
216mm (8.5”)
Nickel-plated zinc Large
0 to 92mm (0 to 3.62”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
229mm (9.02”)
VWR
®
2-Prong Single Adjust Clamps
Material
Size (A) Min. to Max. Grip Size (B) Prong Width (C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Stainless steel
Medium 0 to 75mm (0 to 2.95”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
229mm (9.02”)
Nickle-plated zinc Medium 0 to 75mm (0 to 2.95”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
229mm (9.02”)
Stainless steel
Large
0 to 95mm (0 to 3.74”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
248mm (9.76”)
Nickle-plated zinc Large
0 to 95mm (0 to 3.74”)
23mm (0.91”)
127mm (5”)
11mm (0.43”)
248mm (9.76”)
VWR
®
2-Prong Dual Adjust Clamps
2-Prong Dual Adjust Clamps
2-Prong Single Adjust Clamps
2-Prong Multi-Purpose Clamps
• Stainless steel electro-polished finish or nickel-plated zinc
• Large grip adjustment range
• 2-prong construction
• Single or dual adjust
Designed to securely hold laboratory glassware and apparatus. Extension arm
attaches clamp head securely and offers easy positioning in the deepest fume
hoods. Clamps are constructed with round extension arms, which allow the
clamps to be rotated 360°. Extension arms also allow placement of apparatus
at various distances from lab-frames without compromising the integrity of your
experiment. Stainless steel clamps are electro-polished and made entirely of
stainless steel. They offer exceptional chemical resistance and are autoclavable.
Nickel-plated zinc offers a clamp with a high tensile strength at an economical
price. Supplied with non-slip vinyl sleeves, and for temperatures above 100°C,
fiberglass covers.
NOTE
: An additional holder must be purchased in order to attach clamps to
frames or other apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
Replacement vinyl and fiberglass sleeves are available, page 92.
B
C
E
D
A
(shown in nickel-plated zinc)
(shown in stainless steel)
VWR
®
Talon
®
Multi-Purpose Clamps
VWR
®
CLAMPS
1...,78,79,80,81,82,83,84,85,86,87 89,90,91,92,93,94,95,96,97,98,...116
Powered by FlippingBook