VWR Collection Laboratory Equipment - page 87
85
VWR
®
CLAMPS
VWR
®
3-Prong Multi-Purpose Clamps
• Large grip adjustment range
• 3-prong construction
• Single or dual adjust
• Nickel-plated zinc
Designed to securely hold every type of laboratory glassware and apparatus.
Long, seamless nickle plated brass tubing attaches clamp head securely and
offers easy positioning in the deepest fume hoods. Clamps are constructed with
round extension arms, which allow the clamps to be rotated 360°. Extension
arms also allow placement of apparatus at various distances from lab-frames
without compromising the integrity of your experiment. Supplied with non-slip
vinyl sleeves, and for temperatures above 100°C, fiberglass covers.
NOTE
: An additional holder must be purchased in order to attach clamps to
frames or other apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
Replacement vinyl and fiberglass sleeves are available, page 92.
3-Prong Single Adjust Clamps
3-Prong Dual Adjust Clamps
B
C
E
D
A
Size
(A) Min. to Max. Grip Size
(B) Prong Width (C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Small
0 to 39mm (0 to 1.54”)
11mm (0.43”)
102mm (4”)
8mm (0.32”)
160mm (6.3”)
Medium
0 to 71mm (0 to 2.8”)
19mm (0.75”)
127mm (5”)
11mm (0.43”)
218mm (8.58”)
Large
0 to 108mm (0 to 4.25”)
29mm (1.14”)
127mm (5”)
11mm (0.43”)
248mm (9.76”)
VWR
®
3-Prong Single Adjust Clamps
Size
(A) Min. to Max. Grip Size
(B) Prong Width (C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Small
0 to 46mm (0 to 1.81”)
11mm (0.43”)
102mm (4”)
8mm (0.32”)
168mm (6.61”)
Medium
0 to 69mm (0 to 2.72”)
19mm (0.75”)
127mm (5”)
11mm (0.43”)
229mm (9.02”)
Medium (extended) 0 to 69mm (0 to 2.72”)
19mm (0.75”)
305mm (12”)
11mm (0.43”)
406mm (16”)
Large
0 to 105mm (0 to 4.13”)
29mm (1.14”)
127mm (5”)
11mm (0.43”)
273mm (10.75”)
Large (extended)
0 to 105mm (0 to 4.13”)
29mm (1.14”)
305mm (12”)
11mm (0.43”)
451mm (17.76”)
VWR
®
3-Prong Dual Adjust Clamps
VWR
®
Talon
®
Multi-Purpose Clamps
1...,77,78,79,80,81,82,83,84,85,86 88,89,90,91,92,93,94,95,96,97,...116
Powered by FlippingBook