VWR Collection Laboratory Equipment - page 86
84
Size
(A) Min. to Max. Grip Size
(B) Prong Width
(C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Small
0 to 48mm (0 to 1.9”)
13mm (0.51”)
102mm (4”)
10mm (0.39”)
172mm (6.77”)
Medium 0 to 69mm (0 to 2.72”)
19mm (0.75”)
127mm (5”)
13mm (0.51”)
229mm (9.02”)
Large
0 to 102mm (0 to 4”)
29mm (1.14”)
127mm (5”)
13mm (0.51”)
273mm (10.75”)
VWR
®
3-Prong Stainless Steel Dual Adjust Clamps
B
C
E
D
A
VWR
®
Stainless Steel Multi-Purpose Clamps
• Stainless steel, electro-polished finish
• Large dual adjustment range
• Available in three sizes; small, medium, and large
These durable, multi-purpose clamps are made entirely of stainless steel with
an electro-polished finish and offer excellent chemical resistance and overall
strength. The versatile 3-prong design secures various lab apparatus such
as jointed glassware, columns, flasks, and tubes. Dual prong adjustments
offer a wide range of motion. Long stainless steel extension arm offers easy
positioning and depth adjustment. Autoclavable. Supplied with non-slip vinyl
sleeves, and for temperatures above 100°C, fiberglass covers.
NOTE
: An additional holder must be purchased in order to attach clamps to
frames or other apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
Replacement vinyl and fiberglass sleeves are available, page 92.
Size
Joint Size
Prong Width
Arm Length
Arm Dia.
Overall Length
Cat. No.
Small
24/40mm
16mm (0.63”)
229mm (9.02”)
13mm (0.51”)
356mm (14”)
Large
34/45mm
19mm (0.75”)
229mm (9.02”)
13mm (0.51”)
381mm (15”)
VWR
®
Heavy-Duty Tapered Clamps
Heavy-Duty 4-prong, dual adjust tapered clamps are designed to hold
large vessels with ground glass joint necks. Clamps are constructed
with extension rods for easy attachment to lab-frames and other
apparatus. Nickel-plated zinc construction.
NOTE
: An additional holder must be purchased in order to attach
clamps to frames or other apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
VWR
®
Heavy Duty Tapered Clamps
VWR
®
Talon
®
Multi-Purpose Clamps
VWR
®
CLAMPS
1...,76,77,78,79,80,81,82,83,84,85 87,88,89,90,91,92,93,94,95,96,...116
Powered by FlippingBook