VWR Collection Laboratory Equipment - page 85
83
VWR
®
CLAMPS
VWR
®
Talon
®
Heavy-Duty Clamps
• Large grip adjustment range
• Single or dual adjust
• Available in three sizes; small, medium, and large
• Nickel-plated zinc
VWR
®
patented multi-purpose Talon
®
Heavy-Duty Clamps feature an innovative closed yoke
construction that minimizes contamination and corrosion of internal components. The unique
design enables secure gripping and positioning with added strength and durability.
Available in both single and dual prong adjustment, prongs open gradually to maximize grip
size without binding. Both designs feature precise pressure regulation when gripping glassware
surfaces to reduce the chance of breakage. Talon
®
Clamps are constructed with extension rods
for easy attachment to lab-frames and other apparatus. Supplied with non-slip vinyl sleeves,
and for temperatures above 100°C, fiberglass covers.
The prong height of the Talon
®
Heavy-Duty Clamps make them ideal for use in holding and
securing glassware with precision ground glass joints. The chart details which Talon
®
Clamps
are recommended.
NOTE
: An additional holder must be purchased in order to attach clamps to frames or other
apparatus.
See the Connectors & Holders section of this catalog, pages 93-95.
Replacement vinyl and fiberglass sleeves are available, page 92.
Size
(A) Min. to Max. Grip Size
(B) Prong Width
(C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Small
0 to 32mm (0 to 1.26”)
6mm (0.24”)
102mm (4”)
8mm (0.32”)
152mm (5.98”)
Medium 0 to 70mm (0 to 2.76”)
19mm (0.75”)
127mm (5”)
11mm (0.43”)
222mm (8.74”)
Large
0 to 103mm (0 to 4.06”)
29mm (1.14”)
127mm (5”)
11mm (0.43”)
260mm (10.24”)
VWR
®
3-Prong Dual Adjust Clamps
Size
(A) Min. to Max. Grip Size
(B) Prong Width
(C) Arm Length
(D) Arm Dia.
(E) Overall Length
Cat. No.
Small
0 to 23mm (0 to 0.91”)
6mm (0.24”)
102mm (4”)
8mm (0.32”)
162mm (6.38”)
Medium 0 to 50mm (0 to 1.97”)
19mm (0.75”)
127mm (5”)
11mm (0.43”)
229mm (9.02”)
Large
0 to 72mm (0 to 2.83”)
29mm (1.14”)
127mm (5”)
11mm (0.43”)
260mm (10.24”)
VWR
®
3-Prong Single Adjust Clamps
B
C
E
D
A
Ground Glass Joint Reference Chart
Clamp
Joint Size (mm)
Small Dual Adjust
10/30, 12/30, 14/20, 19/22, 24/40, 29/42, 34/45
Medium Dual Adjust
14/40, 19/38
Large Dual Adjust
45/50
Single Adjust Clamps
Dual Adjust Clamps
VWR
®
Talon
®
Multi-Purpose Clamps
1...,75,76,77,78,79,80,81,82,83,84 86,87,88,89,90,91,92,93,94,95,...116
Powered by FlippingBook