VWR Collection Laboratory Equipment - page 83
81
24-Well 4mL Vial Sets, polycarbonate
Plastic case with foam rack holds 24, 4mL polycarbonate vials. Each vial includes one,
3/8” (1cm) stainless steel grinding ball and threaded lid. Lid is available unlined or with
lining for added secure closure. Sold as a case of 10 sets (240 vials).
24-Well 4mL Vial Sets, polyethylene
Plastic case with foam rack holds 24, 4mL polyethylene vials. Each vial includes one,
3/8” (1cm) stainless steel grinding ball and threaded lid. Lid is available unlined or with
lining for added secure closure. Sold as a case of 10 sets (240 vials).
5-Well 15mL Vial Set, polycarbonate
Plastic case with foam rack holds 5, 15mL polycarbonate vials for larger samples. Each
vial includes two, 7/16” (1.1cm) stainless steel grinding balls and threaded lid. Sold as
a case of 10 sets (50 vials).
24-Pin Magnet and Magnet Stand/Ball Separator
The 24-Pin Magnet can be used to remove grinding balls before samples are added to
the vials and then, with the use of the Magnet Stand/Ball Separator, used to align and
drop grinding balls into vials. Each 2” (5.1cm) pin is capped with a strong neodymium
rare earth magnet. The pins of this magnet can also be used to remove smaller grinding
balls from 96-well plates.
Ball Dropper/Bead Dispenser
Ball Dropper/Bead Dispenser can be used to add 5/32” stainless steel balls to deep well
plates, and to dispense small grinding beads into empty micro-well plates. Includes ball
dropper, reservoir tray, grinding bead swipe card, and plastic retaining sheet.
Magnet Pipette Tips
Magnet Pipette Tips are a simple, cost effective solution for removing stainless steel
grinding balls from 96-well plates and grinding vials. The tips are made to fit most
0-200µl pipettes and work with steel and 440C stainless steel balls.
Description
Cat. No.
Magnet Pipette Tips, pack of 12
Description
Lid
Cat. No.
24-Well 4mL Vial Set, Polyethylene
Unlined
24-Well 4mL Vial Set, Polyethylene
Lined
Description
Lid
Cat. No.
24-Well 4mL Vial Set, Polycarbonate
Unlined
24-Well 4mL Vial Set, Polycarbonate
Lined
Description
Lid
Cat. No.
5-Well 15mL Vial Set, Polycarbonate
Unlined
Description
Cat. No.
24-Pin Magnet
Magnet Stand/Ball Separator
Description
Cat. No.
Ball Dropper/Bead Dispenser
VWR
®
High Throughput Homogenizer Accessories
VWR
®
HOMOGENIZERS
1...,73,74,75,76,77,78,79,80,81,82 84,85,86,87,88,89,90,91,92,93,...116
Powered by FlippingBook