VWR Collection Laboratory Equipment - page 82
80
Replacement Accessories
VWR
®
High Throughput Homogenizer
Description
Electrical (50/60 Hz)
Cat. No.
High Throughput Homogenizer
120V | 6.3 amps | 450 watts
High Throughput Homogenizer
230V | 3.1 amps | 450 watts
• Ability to process small quantities
• Eliminates cross contamination
• Rapid sample processing
The VWR
®
High Throughput Homogenizer is specifically designed for high throughput
sample processing in a microplate, vial set format or any 4 x 5 x 2.5 matrix to produce
effective homogenization of tissue samples. Homogenizer produces a reciprocal motion
that is optimized for grinding difficult samples. Easy-to-load tray securely locks microplate
or sample holder in place. The high speed linear motion of the homogenizer allows for
rapid sample processing; in most cases two minutes or less. Analog controls.
Homogenized samples can be subsequently handled manually or in an automated
system. The High Throughput Homogenizer is suitable for the isolation of protein, DNA,
RNA, viruses, and other biological components released during homogenization.
Animal tissue, seeds, tubers, leaf punches, soil and sediment samples, insects, and
microbial cultures can all be effectively homogenized in a 96-well or vial set format. The
unit is also useful for pulverizing dry samples for chemical analysis and solubility studies.
Unit includes a 92” (234cm), 3-wire cord and plug (230 volt units are supplied with Euro
type plug). 2 year limited warranty on parts and labor.
Grinding Accessories
Grinding Vial Sets
combine the benefits of the microplate format and the size of vials
to provide a tool for higher throughput sample grinding. Vials are available in either
polycarbonate or polyethylene materials. The polyethylene vial set is offered in a 24 vial,
4mL size. The polycarbonate vial set is offered in two sizes; 24 vial, 4mL size and 5 vial,
15mL size. The 4mL vial size is conveniently packaged as a set or in bulk.
Grinding balls
are used with deep well plates and vial sets to bead beat plant tissues,
seeds, and animal tissues. These grinding balls are made of 440C stainless steel which
allows them to be retrieved with a magnet while making them resistant to tarnishing.
Grinding balls are treated to remove residual oils and contaminants prior to packaging.
Recommended Applications:
3/8” Good for homogenizing seeds and larger samples of animal tissue in 4mL vials.
7/16” Suitable for larger grinding jobs, such as pooled seeds, in 15 mL vials.
5/32” Use with deep well plates for leaf punches or animal tissue of 50mg or less.
When homogenization is complete, grinding balls can be removed with optional 24-Pin
Magnet.
Operating Conditions:
Units can be run in conditions from 5 to 40°C (41 to 104°F), 20% to 85% relative humidity,
non-condensing.
Specifications
Speed Range
0 to 1600rpm
Timer
1 to 10 minutes
Maximum Capacity
1 deep well plate, 4 stacked standard well
plates, or 1 vial set; 300 gram maximum
Reciprocal Stroke
1.2” (3.1cm)
Dimensions, Overall (L x W x H)
17.5 x 15 x 14.2”
(44.5 x 38.1 x 36.1cm)
Shipping Weight
75lbs (34.1kg)
Description
Lid
Cat. No.
Bulk 4mL Polyethylene Vials, case of 240
Unlined
Bulk 4mL Polyethylene Vials, case of 240
Lined
Bulk 4mL Polycarbonate Vials, case of 240
Unlined
Bulk 4mL Polycarbonate Vials, case of 240
Lined
3/8” Stainless Steel Grinding Balls, pack of 1000
N/A
5/32” Stainless Steel Grinding Balls, pack of 5000
N/A
corn
rice
soy beans
VWR
®
High Throughput Homogenizer
VWR
®
HOMOGENIZERS
1...,72,73,74,75,76,77,78,79,80,81 83,84,85,86,87,88,89,90,91,92,...116
Powered by FlippingBook