VWR Collection Laboratory Equipment - page 80
78
Foam Test Tube Racks
Description
Tube Capacity
Color
Dimensions (L x W x H)
Cat. No.
10mm Test Tube Foam Rack
50
Gray
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
12mm Test Tube Foam Rack
50
Blue
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
13mm Test Tube Foam Rack
50
Yellow
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
16mm Test Tube Foam Rack (for 15mL centrifuge tubes)
50
Green
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
25mm Test Tube Foam Rack
28
White
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
29mm Test Tube Foam Rack (for 50mL centrifuge tubes)
15
Red
5.5 x 9.5 x 2” (14 x 24.1 x 5.1cm)
Replacement Tray Pad Set (upper & lower)
N/A
Gray
7 x 12 x 1” (17.8 x 30.5 x 2.5cm)
Post Extension Kit
Adds 6” (15.2cm) to the post of the Multi-Tube Vortex Mixer to accommodate tubes up to 10” (25.4cm) tall.
VWR
®
Foam Test Tube Racks
Description
Cat. No.
Post Extension Kit
VWR
®
Post Extension Kit
VWR
®
Multi-Tube Vortexer Accessories
VWR
®
VORTEX MIXERS
1...,70,71,72,73,74,75,76,77,78,79 81,82,83,84,85,86,87,88,89,90,...116
Powered by FlippingBook