VWR Collection Laboratory Equipment - page 8
6
VWR
®
SHAKERS
Model
Orbital Shaker
Standard 3500
Orbital Shaker
Advanced 3500
Reciprocating Shaker
Advanced 3750
Orbital Shaker
Standard 5000
Speed Range
25 to 500rpm
15 to 500rpm
20 to 300rpm
25 to 500rpm
Timer
1 to 120 minutes
1 second to 160 hours
1 second to 160 hours
1 to 120 minutes
Motion
Orbital
Orbital
Reciprocating
Orbital
Orbit
19mm
19mm
Stroke: 19mm
25mm
Maximum Weight Capacity
35lbs
35lbs
15lbs
50lbs
Audible Alarm
NO
YES
YES
NO
Load Sensor
NO
YES
YES
NO
Motor Type
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
CO
2
Environment
YES
YES
YES
YES
Overload Protection
NO
YES
YES
NO
User Calibration (Speed)
NO
YES
YES
NO
RS-232 Interface
NO
YES
YES
NO
Included Tray (L x W)
11 x 13”
11 x 13”
11 x 13”
18 x 24”
Tray / Platform Options (L x W)
11 x 13”, 13 x 13”, 18 x 18”, 18 x 24”
Adjustable Platform
Separatory Funnel Platform
11 x 13”, 13 x 13”, 18 x 18”, 18 x 24”
Adjustable Platform
Separatory Funnel Platform
11 x 13”, 13 x 13”, 18 x 18”, 18 x 24”
Adjustable Platform
Separatory Funnel Platform
18 x 24”, 18 x 30”
Adjustable Platform
Large Vessel Carrier Platform
Model
Orbital Shaker
Standard 1000
Mini
Shaker
Mini
Shaker 15
Microplate
Shaker
Speed Range
40 to 300rpm
100 to 1200rpm
40 to 300rpm
100 to 1200rpm
Timer
N/A
1 second to 160 hours
1 second to 160 hours
1 second to 160 hours
Motion
Orbital
Orbital
Orbital
Orbital
Orbit
15mm
3mm
15mm
3mm
Maximum Weight Capacity
8lbs
8lbs
8lbs
4 microplates / 2 micro-tube racks
Audible Alarm
NO
YES
YES
YES
Load Sensor
NO
NO
NO
NO
Motor Type
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
Brushless DC Motor
CO
2
Environment
YES
YES
YES
YES
Overload Protection
NO
YES
YES
YES
User Calibration (Speed)
NO
NO
NO
NO
RS-232 Interface
NO
NO
NO
NO
Included Tray (L x W)
11.75 x 8.75”
11.75 x 8.75”
11.75 x 8.75”
11 x 7.75”
Tray / Platform Options (L x W)
N/A
Adjustable Platform
Adjustable Platform
N/A
VWR
®
Open-Air Shaker Selection Guide
1,2,3,4,5,6,7 9,10,11,12,13,14,15,16,17,18,...116
Powered by FlippingBook