VWR Collection Laboratory Equipment - page 78
76
• Process up to 50 samples at a time
• Continuous or timed operation
Conveniently angled front panel features rocker switch for run, time, and
standby modes.
Operating Features:
Microprocessor control:
The variable speed microprocessor control
provides consistent uniform mixing action.
Adjustment knobs:
Basic speed and time knobs with 1 to 10 dial settings.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85%
relative humidity, non-condensing.
Applications:
Suspensions, high throughput testing in clinical, environmental, and chemistry
labs.
Ordering Information
Unit includes a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Unit is also supplied with a Tray Pad Set for
support and one, 12mm Test Tube Foam Rack (
2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Standard Multi-Tube Vortexer
120V | 0.8 amps | 100 watts
Standard Multi-Tube Vortexer
230V | 0.4 amps | 100 watts
VWR
®
Standard Multi-Tube Vortexer
Speed Range*
1200 to 2400rpm
Timer
0 to 60 seconds
Orbit
3.6mm (0.14”)
Maximum Weight Capacity
10lbs (4.5kg)
Duty Rating
Continuous duty
Tray Dimensions (L x W)
7.25 x
12.25” (18.4 x 31.1cm)
Overall Dimensions (L x W x H)
9.5 x
15.1 x 16”
( 24.1 x 38.4 x 40.6 cm)
Ship Weight
42lbs (19.1kg)
Specifications
* Maximum speed will vary depending on load.
VWR
®
Standard Multi-Tube Vortexers
VWR
®
VORTEX MIXERS
1...,68,69,70,71,72,73,74,75,76,77 79,80,81,82,83,84,85,86,87,88,...116
Powered by FlippingBook