VWR Collection Laboratory Equipment - page 75
73
VWR
®
Analog Vortex Mixer
• Variable, analog speed control
• Continuous or touch mode operation
Control allows low rpm startup for gentle shaking or high speed mixing for vigorous
vortexing of samples. Two modes of operation; continuous mode when using accessory
attachments or touch mode which activates mixing when depressing the cup head.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative
humidity, non-condensing.
Ordering Information:
Includes both cup head and three-inch head with cover. 2 year limited warranty on
parts and labor.
Speed Range* 1
20V
300 to 3200rpm
230V
300 to 2500rpm
Orbit
4.9mm
(
0.194”)
Duty Rating
Intermittent Duty
Controls
Auto/Off/On Rocker Switch,
Speed Knob: Variable 1 to 10 Dial Marks
Dimensions (L x W x H)
7.75 x 6 x 5.5 (19.69 x 15.24 x 13.97cm)
Ship Weight
11.7lbs (5.3kg)
Specifications
Description
Electrical (50/60 Hz)
Cat. No.
Analog Vortex Mixer
120V | 1.2 amps | 150 watts
Analog Vortex Mixer
230V | 0.6 amps | 150 watts
VWR
®
Analog Vortex Mixer
VWR
®
Pulsing Vortex Mixer
• LED displays for time and speed
• Continuous or touch mode operation
• Microprocessor controls
• Glass bead cell disruption/homogenization
Powerful pulsing vortex action produces excellent cell disruption for glass bead
procedures. Capable of complete cell disruption of samples in only minutes. Unique
pulsing action reduces heat generation while providing more effective mixing and
disruption. Displayed time counts up during continuous operation and counts down
during timed runs.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative
humidity, non-condensing.
Ordering Information:
Includes cup head, three inch head with cover, and an easy-to-load 1.5mL to 2mL
Micro-Tube Holder. Holder has a built-in cup head. 2 year limited warranty on parts
and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Pulsing Vortex Mixer
120V | 1.2 amps | 150 watts
Pulsing Vortex Mixer
230V | 0.6 amps | 150 watts
Speed Range*
120V
500 to 3000rpm
230V
500 to 2500rpm
Timer
1 second to 160 hours
Orbit
2.5mm (0.098”)
Duty Rating
Intermittent Duty
Controls
Auto/Standby/On Rocker Switch,
LED Displays For Speed/Time,
Up/Down Buttons For Set-Point Control
Pulse Button
Dimensions (L x W x H)
7.75 x 6 x 5.5 (19.69 x 15.24 x 13.97cm)
Ship Weight
11.7lbs (5.3kg)
VWR
®
Pulsing Vortex Mixer
Specifications
VWR
®
Analog & Pulsing Vortex Mixers
VWR
®
VORTEX MIXERS
* Maximum speed will vary depending on accessory used.
* Maximum speed will vary depending on accessory used.
1...,65,66,67,68,69,70,71,72,73,74 76,77,78,79,80,81,82,83,84,85,...116
Powered by FlippingBook