VWR Collection Laboratory Equipment - page 74
72
VWR
®
Fixed Speed Vortex Mixer
• Fixed high speed mixing
• Touch mode operation
Starts mixing when the cup head is pressed down. Speed is fixed at full rpm to
provide vigorous vortexing of samples.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative
humidity, non-condensing.
Ordering Information:
Includes both cup head and three-inch head with cover. 2 year limited warranty on
parts and labor.
Speed Range*
120V
3200rpm
230V
2500rpm
Orbit
4.9mm
(
0.194”)
Controls
None
Duty Rating
Intermittent Duty
Dimensions (L x W x H)
7.75 x 6 x 5.5 (19.69 x 15.24 x 13.97cm)
Ship Weight
11.7lbs (5.3kg)
Description
Electrical (50/60 Hz)
Cat. No.
Fixed Speed Vortex Mixer
120V | 1.2 amps | 150 watts
Fixed Speed Vortex Mixer
230V | 0.6 amps | 150 watts
* Maximum speed will vary depending on accessory used.
Specifications
VWR
®
Fixed Speed Vortex Mixer
VWR
®
Fixed Speed & Digital Vortex Mixers
VWR
®
VORTEX MIXERS
VWR
®
Digital Vortex Mixer
• LED displays for speed and time
• Continuous or touch mode operation
• Microprocessor controls
Ideal for applications that demand repeatable results. Microprocessor control maintains
set speed for strong, consistent mixing action. Timer will display elapsed time or,
when programmed to user defined time limit, the unit will shut off when time reaches
zero. Two modes of operation; continuous mode when using accessory attachments
or touch mode which activates mixing when depressing the cup head.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative
humidity, non-condensing.
Ordering Information:
Includes both cup head and three-inch head with cover. 2 year limited warranty on
parts and labor.
Speed Range*
120V
500 to 3000rpm
230V
500 to 2500rpm
Timer
1 second to 160 hours
Orbit
4.9mm
(
0.194”)
Duty Rating
Intermittent Duty
Controls
Auto/Standby/On Rocker Switch,
LED Displays For Speed/Time,
Up/Down Buttons For Set-Point Control
Dimensions (L x W x H)
7.75 x 6 x 5.5 (19.69 x 15.24 x 13.97cm)
Ship Weight
11.7lbs (5.3kg)
Description
Electrical (50/60 Hz)
Cat. No.
Digital Vortex Mixer
120V | 1.2 amps | 150 watts
Digital Vortex Mixer
230V | 0.6 amps | 150 watts
Specifications
VWR
®
Digital Vortex Mixer
* Maximum speed will vary depending on accessory used.
1...,64,65,66,67,68,69,70,71,72,73 75,76,77,78,79,80,81,82,83,84,...116
Powered by FlippingBook