VWR Collection Laboratory Equipment - page 73
71
Microplate Holder (single)
Designed to hold one standard microplate
Cat. No.
Foam Insert for 15-18mm test tubes
Foam insert holds (20) 15-18mm diameter test tubes. Ideal
for 15mL centrifuge tubes. Requires Universal Holder.
Cat. No.
15-18mm test tubes
Universal Holder
Foam Insert for 12-13mm test tubes
Foam insert holds (34) 12-13mm diameter test tubes.
Requires Universal Holder.
Cat. No.
12-13mm test tubes
Universal Holder
Foam Insert for 0.5mL microtubes
Foam insert holds (52) 0.5mL microtubes.
Requires Universal Holder.
Cat. No.
0.5mL microtubes
Universal Holder
Foam Insert for 1.5 to 2.0mL microtubes
Foam insert holds (38) 1.5 to 2.0mL microtubes.
Requires Universal Holder.
Cat. No.
1.5 to 2.0mL microtubes
Universal Holder
Microplate Holder (double)
Designed to hold two standard microplates.
Cat. No.
Flat Foam Insert
Ideal for custom applications. Can be cut or drilled
to fit your specifications. Requires Universal Holder.
Cat. No.
Flat Foam Insert
Universal Holder
Foam Insert for 19-21mm test tubes
Foam insert holds (18) 19-21mm diameter test tubes.
Requires Universal Holder.
Cat. No.
19-21mm test tubes
Universal Holder
Foam Insert for 22-25mm test tubes
Foam insert holds (13) 22-25mm diameter test tubes.
Requires Universal Holder.
Cat. No.
22-25mm test tubes
Universal Holder
Foam Insert for 26-29mm test tubes
Foam insert holds (4) 26-29mm diameter test tubes. Ideal
for 50mL centrifuge tubes. Requires Universal Holder.
Cat. No.
26-29mm test tubes
Universal Holder
Small Vessel Holder
Rubber holder secures 125 and 250mL Erlenmeyer
flasks. Vessel holder also includes a grip mat.
Requires Universal Holder.
Cat. No.
Small Vessel Holder
Universal Holder
Stackable Microplate Holder Four
Designed to maximize the capacity of the Heavy-Duty
Vortex Mixer to eight microplates by stacking the tray
on top of the four plate holder.
Cat. No.
Microplate Holder (quad)
Designed to hold four standard microplates.
Cat. No.
Single Tube Holder
Single tube hands free mixing designed to fit on the Heavy-
Duty Vortex Mixer. Easily attached to the top of mixer and is
magnetically secured. Accepts tubes from 2.5 to 4.5” (6.4-
11.4cm). Minimum tube diameter is 0.75” (19mm).
Cat. No.
Universal Holder & Cover
Replacement for items supplied with Heavy-Duty
Vortex Mixer. Cover allows for mixing irregularly
shaped objects.
Cat. No.
Universal Holder
Universal Holder Cover
Cup Head
Designed for mixing 1 tube at a time.
Cat. No.
Large Vessel Holder
Rubber holder secures 500 and 1000mL Erlenmeyer
flasks. Vessel holder also includes a grip mat.
Requires Universal Holder.
Cat. No.
Large Vessel Holder
Universal Holder
VWR
®
Accessories
VWR
®
VORTEX MIXERS
1...,63,64,65,66,67,68,69,70,71,72 74,75,76,77,78,79,80,81,82,83,...116
Powered by FlippingBook