VWR Collection Laboratory Equipment - page 72
70
Specifications
Specifications
VWR
®
Standard Heavy-Duty Vortex Mixer
• Designed for continuous duty
The VWR
®
Standard Heavy-Duty Vortex Mixer is a variable speed analog mixer that is
designed for continuous duty. The heavy-duty design and efficient motor allow this mixer
to operate in continuous duty and handle all accessories over the entire speed range.
Choose from two modes of operation: “Touch” mode for mixing tubes when cup head or
Universal Holder with cover is depressed or “On” mode when using any of the accessory
attachments for continuous operation.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative humidity,
non-condensing.
Ordering Information:
Mixer includes a cup head, Universal Holder with cover, foam microtube insert for (38) 1.5
to 2.0 mL microtubes, and a 92” (234cm) detachable, 3-wire cord and plug (230 volt units
are supplied with Euro type plug). 2-year limited warranty on parts and labor.
Speed Range
On Mode:
300-2500rpm
Touch Mode:
300-3500rpm
Orbit
4.9mm (0.194”)
Controls
Analog
Capacity
2.5lbs (1.1kg)
Duty Rating
Continuous duty
Dimensions (L x W x H)
9.5 x 6.6 x 6.3” (24.1 x 16.8 x 16cm)
Ship Weight
15lbs (6.8kg)
Description
Electrical (50/60 Hz)
Cat. No.
Standard Vortex Mixer
120V | 0.25 amps | 30 watts
Standard Vortex Mixer Euro Plug 230V | 0.13 amps | 30 watts
VWR
®
Advanced Heavy-Duty Vortex Mixer
• Designed for continuous duty
• LED displays for speed and time
The VWR
®
Advanced Heavy-Duty Vortex Mixer is ideal for applications that demand
repeatable results. Mixer features touchpad controls and LED displays for accurate speed
(rpm) and time (minutes and seconds) results. The heavy-duty design and efficient motor
allow this mixer to operate in continuous duty and handle all accessories over the entire
speed range. Choose from two modes of operation: “Touch” mode for mixing tubes when
cup head or Universal Holder with cover is depressed or “On” mode, when using any of
the accessory attachments for continuous operation. Microprocessor control maintains
set speed for strong, consistent mixing action. Timer will display elapsed time or, when
programmed to user defined time limits, the unit will shut off when time reaches zero.
Ordering Information:
Mixer includes a cup head, Universal Holder with cover, foam microtube insert for (38) 1.5
to 2.0 mL microtubes, and a 92” (234cm) detachable, 3-wire cord and plug (230 volt units
are supplied with Euro type plug). 2-year limited warranty on parts and labor.
Speed Range
On Mode:
300-2500rpm
Touch Mode:
300-3500rpm
Timer
1 second to 160 hours
Orbit
4.9mm (0.194”)
Controls
Touch/Standby/On Rocker Switch,
LED Displays For Speed/Time,
Up/Down Buttons For Set-Point Control
Capacity
2.5lbs (1.1kg)
Duty Rating
Continuous duty
Dimensions (L x W x H)
9.5 x 6.6 x 6.3” (24.1 x 16.8 x 16cm)
Ship Weight
15lbs (6.8kg)
Description
Electrical (50/60 Hz)
Cat. No.
Advanced Vortex Mixer
120V | 0.25 amps | 30 watts
Advanced Vortex Mixer Euro Plug 230V | 0.13 amps | 30 watts
VWR
®
Advanced Heavy-Duty Vortex Mixer
VWR
®
Standard Heavy-Duty Vortex Mixer
VWR
®
Standard & Advanced Heavy-Duty Vortex Mixers
VWR
®
VORTEX MIXERS
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85%
relative humidity, non-condensing.
with
optional insert
with
optional microplate
holders
and
1...,62,63,64,65,66,67,68,69,70,71 73,74,75,76,77,78,79,80,81,82,...116
Powered by FlippingBook