VWR Collection Laboratory Equipment - page 71
69
VWR
®
Advanced Microplate Vortex Mixer
VWR
®
Standard Microplate Vortex Mixer
• Designed for continuous duty
• Designed for shaking microplates or tubes
The VWR
®
Standard Microplate Vortex Mixer is a variable speed analog mixer that is
designed for continuous duty. The high speed and small orbit is optimal for effectively
mixing microplates. Choose from two modes of operation: “Touch” mode which activates
mixing when cup head is depressed or “On” mode when using any of the accessory
attachments for continuous operation.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative humidity,
non-condensing.
Ordering Information:
Mixer includes a cup head, single microplate holder, and a 92” (234cm) detachable, 3-wire
cord and plug. (230 volt units are supplied with Euro type plug). Accessories that can be
used on the Microplate Vortex Mixer are cup head, single microplate holder and double
microplate holder. 2-year limited warranty on parts and labor.
VWR
®
Advanced Microplate Vortex Mixer
• Designed for continuous duty
• Designed for shaking microplates or tubes
• LED displays for speed and time
The VWR
®
Advanced Microplate Vortex Mixer is ideal for applications that demand
repeatable results. Mixer features touchpad controls and LED displays for accurate speed
(rpm) and time (minutes and seconds) results. Microprocessor control maintains set speed
for strong, consistent mixing action. Timer will display elapsed time or when programmed
to a user defined time limit, the unit will shut off when time reaches zero. Choose from two
modes of operation: “Touch” mode which activates mixing when cup head is depressed
or “On” mode when using any of the accessory attachments for continuous operation.
Operating Conditions:
Unit can be run in conditions from 4 to 40°C (39 to 104°F), 20% to 85% relative humidity,
non-condensing.
Ordering Information:
Mixer includes a cup head, single microplate holder, and a 92” (234cm) detachable, 3-wire
cord and plug. (230 volt units are supplied with Euro type plug). Accessories that can be
used on the Microplate Vortex Mixer are cup head, single microplate holder and double
microplate holder. 2-year limited warranty on parts and labor.
Speed Range
On Mode:
300-2500rpm
Touch Mode:
300-3500rpm
Timer
1 second to 160 hours
Orbit
3.5mm (0.13”)
Controls
Touch/Standby/On Rocker Switch,
LED Displays For Speed/Time,
Up/Down Buttons For Set-Point Control
Capacity
2 microplates
Duty Rating
Continuous duty
Dimensions (L x W x H)
10.5 x 5.4 x 4.5”
(26.7 x 13.7 x 11.4cm)
Ship Weight
12lbs (5.4kg)
Description
Electrical (50/60 Hz)
Cat. No.
Advanced Vortex Mixer
120V | 0.25 amps | 30 watts
Advanced Vortex Mixer Euro Plug 230V | 0.13 amps | 30 watts
Specifications
Speed Range
On Mode:
300-2500rpm
Touch Mode:
300-3500rpm
Orbit
3.5mm (0.13”)
Controls
Analog
Capacity
2 microplates
Duty Rating
Continuous duty
Dimensions (L x W x H)
10.5 x 5.4 x 4.5” (26.7 x 13.7 x 11.4cm)
Ship Weight
12lbs (5.4kg)
Specifications
Description
Electrical (50/60 Hz)
Cat. No.
Standard Vortex Mixer
120V | 0.25 amps | 30 watts
Standard Vortex Mixer Euro Plug 230V | 0.13 amps | 30 watts
VWR
®
Standard Microplate Vortex Mixer
VWR
®
Standard & Advanced Microplate Vortex Mixers
VWR
®
VORTEX MIXERS
1...,61,62,63,64,65,66,67,68,69,70 72,73,74,75,76,77,78,79,80,81,...116
Powered by FlippingBook