VWR Collection Laboratory Equipment - page 70
68
Solid Blocks
Single and double block. For use as a low-temperature hotplate, slide drying, or for custom drilling to
make a custom block.
Block Size
Dimension (L x W x H)
Cat. No.
Single
3.75 x 3 x 2” (9.5 x 7.6 x 5.1cm)
Double
6 x 3.75 x 2.25” (15.2 x 9.5 x 5.7cm)
Stainless Steel Sand Baths
Ideal for irregular vessels. Stainless steel construction for superior corrosion resistance. Designed to
hold sand, stainless steel shot or non-volatile liquids.
For Unit
Dimensions (L x W x H)
Cat. No.
1 Block Dry Block Heater
3.75 x 3 x 2.5” (9.5 x 7.6 x 6.4cm)
2 Block Dry Block Heater
3.75 x 5.9 x 2.5” (9.5 x 14.9 x 6.4cm)
3 Block Dry Block Heater
3.75 x 8.8 x 2.5” (9.5 x 22.4 x 6.4cm)
4 Block Dry Block Heater
5.8 x 7.5 x 2.5” (14.7 x 19.1 x 6.4cm)
6 Block Dry Block Heater
5.8 x 11.3 x 2.5” (14.7 x 28.7 x 6.4cm)
Accessory
Cat. No.
Sand, 1lb (0.454kg)
Stainless Steel Shot, 1lb (0.454kg)
Low Temperature Covers
Plexiglass cover reduces air flow for additional temperature stability in low-temperature applications. Two
sides are open 0.25” (6.4mm).
Block Size
Dimensions (L x W x H)
Cat. No.
2 Block Cover
6.5 x 6.5 x 1.6” (16.5 x 16.5 x 4.1cm)
4 Block Cover
8.5 x 8.5 x 1.6” (21.6 x 21.6 x 4.1cm)
6 Block Cover
12.5 x 8.5 x 1.6” (31.8 x 21.6 x 4.1cm)
VWR
®
Modular Blocks & Accessories
VWR
®
DRY BLOCK HEATERS
1...,60,61,62,63,64,65,66,67,68,69 71,72,73,74,75,76,77,78,79,80,...116
Powered by FlippingBook