VWR Collection Laboratory Equipment - page 7
5
VWR
®
SHAKERS
VWR
®
Thermal Shake Touch Modular Blocks
VWR
®
Microplate Block
Sample Type
No. of Wells
Well Size
Well Depth Dimensions
Cat. No.
Microplate Thermal Block with Lid
1
5.06 x 3.38” (12.9 x 8.6cm) 1.00” (2.5cm) 6.5 x 4.75 x 4” (15.2 x 10.2 x 5.1cm)
VWR
®
Tube Blocks
Sample Type
No. of Wells
Well Diameter
Well Depth Dimensions
Cat. No.
0.5mL Tubes*
30
0.312” (0.79cm)
1.01” (2.57cm) 5.7 x 4.1 x 1.8” (14.5 x 10.4 x 4.6cm)
1.5mL Tubes*
24
0.438” (1.11cm)
1.32” (3.35cm) 5.7 x 4.1 x 2.1” (14.5 x 10.4 x 5.3cm)
2.0mL Tubes*
24
0.454” (1.15cm)
1.32” (3.35cm) 5.7 x 4.1 x 2.1” (14.5 x 10.4 x 5.3cm)
5-7mL Tubes
24
0.472” (1.20cm)
1.35” (3.43cm) 5.7 x 4.1 x 2.2” (14.5 x 10.4 x 5.6cm)
VWR
®
Cryo Tube Block
Sample Type
No. of Wells
Well Diameter
Well Depth Dimensions
Cat. No.
2.0mL Cryo Tubes
24
0.496” (1.26cm)
1.34” (3.40cm) 5.7 x 4.1 x 2.2” (14.5 x 10.4 x 5.6cm)
VWR
®
Conical Tube Blocks
Sample Type
No. of Wells
Well Diameter
Well Depth Dimensions
Cat. No.
15mL Conical Tubes
9
0.680” (1.73cm)
4.02” (10.2cm) 5 x 4 x 5” (12.7 x 10.2 x 12.7cm)
50mL Conical Tubes
4
1.180” (3.00cm)
3.89” (9.88cm) 5 x 4 x 5” (12.7 x 10.2 x 12.7cm)
* Supplied with rack and cover
1,2,3,4,5,6 8,9,10,11,12,13,14,15,16,17,...116
Powered by FlippingBook