VWR Collection Laboratory Equipment - page 69
67
Combination Blocks
Single block. These blocks have been designed for variable sized samples.
Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
Test Tube Combination:
6mm
6
8.3mm 48.4mm
12/13mm 5
13.9mm 48.4mm
25mm
3
26.2mm 48.4mm
Centrifuge Tube Combination: 1.5mL
4
11.1mm 39.1mm
15mL
3
17.1mm 44.5mm
50mL
2
29mm 47.6mm
Micro-Tube Combination:
0.5mL
6
7.9mm 27.6mm
1.5mL
10
11.1mm 39.1mm
2mL
5
11.5mm 38.1mm
Vial Blocks
Single block. Designed for sample/serum and scintillation vials.
Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
12mm Vial
20
12.7mm 30mm
15mm Vial
20
15.8mm 35mm
16mm Vial
15
16.4mm 45mm
17mm Vial
12
17.8mm 45mm
19mm Vial
12
19.7mm 45mm
21mm Vial
9
21.7mm 45mm
23mm Vial
8
23.8mm 45mm
25mm Vial
8
25.8mm 45mm
28mm Vial
6
28.8mm 45mm
PCR - Plate, Tube, Strip Blocks
Single and double block. Tapered tube wells for 0.2mL tubes. Spaced for easy access and removal.
Sample Type
Block Size No. of Wells Well Dia.
Well Depth
Cat. No.
96-well PCR Plate
Double
96
6.4mm 15.5mm
10 x 8 Tube Strips
Single
80
6.4mm 15.5mm
Individual Tubes
Single
64
6.4mm 20.2mm
Cuvette Block
Single block. Two parallel slots fit 6 cuvettes in each slot, side-by-side.
Sample Type
No. of Wells
Well Depth
Cat. No.
(12) 12.5mm Cuvettes
2
25.4mm
VWR
®
Modular Blocks & Accessories
VWR
®
DRY BLOCK HEATERS
1...,59,60,61,62,63,64,65,66,67,68 70,71,72,73,74,75,76,77,78,79,...116
Powered by FlippingBook