VWR Collection Laboratory Equipment - page 68
66
VWR
®
Modular blocks are constructed from a solid anodized aluminum block. The close contact of tubes-to-block-walls allow for maximum heat retention.
Each block has a thermometer well for measuring block temperature.
Caution!
To avoid possible electrical hazard, do not fill well or block with water or
other liquids. Units are designed as a dry bath/incubator.
Single block dimensions (L x W x H):
3.75 x 3 x 2” (9.5 x 7.6 x 5.1cm),
Double block dimensions (L x W x H):
6 x 3.75 x 2.25” (15.2 x 9.5 x 5.7cm)
Microcentrifuge Tube Blocks
Single block.
Brand/Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
0.5mL Tube
30
7.9mm
27.6mm
1.5mL Tube
20
11.1mm
39.1mm
1.5mL Eppendorf™ Tube
20
11.5mm
36.9mm
2mL Eppendorf™ Tube
20
11.5mm
38.1mm
2mL Corning™ Tube
20
10.9mm
38.1mm
Titer Plate Block
Double block. Fits 2/4/6 block Dry Block Heaters. Ideal for 96-well or 384-well titer plates. Recessed well
for better stability, flat surface good for flat and round bottom plates.
Sample Type
Well Depth
Cat. No.
Titer Plate
13.5mm
Conical-Bottom Centrifuge Tube Blocks
Single block.
Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
15mL Tube
12
17.1mm
47.2mm
50mL Tube
5
29mm
47.6mm
Standard Test Tube Blocks
Single block.
Sample Type
No. of Wells Well Dia.
Well Depth
Cat. No.
6mm Tube
30
8.3mm
48.4mm
10mm Tube
24
10.7mm
48.4mm
12/13mm Tube
20
13.9mm
48.4mm
12/13mm Tube
16
13.9mm
48.4mm
15/16mm Tube
12
17.5mm
48.4mm
17/18mm Tube
12
19.1mm
48.4mm
20mm Tube
8
21mm
48.4mm
25mm Tube
6
26.2mm
48.4mm
35mm Tube
4
35mm
47.6mm
VWR
®
Modular Blocks & Accessories
VWR
®
DRY BLOCK HEATERS
1...,58,59,60,61,62,63,64,65,66,67 69,70,71,72,73,74,75,76,77,78,...116
Powered by FlippingBook