VWR Collection Laboratory Equipment - page 65
63
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
Modular blocks are sold separately (see pages 66-68).
External Temperature Probe Kit
Enables the unit to read actual block or sample temperature and display that
temperature on the control panel. The optional External Temperature Probe Kit
includes a stainless steel RTD PT1000 probe, 18” (45.7cm) stainless steel support
rod, thermometer / temperature probe extension clamp and hook connector. The
RTD probe is designed to fit perfectly in to the thermometer well of each modular
block.
Description
Cat. No.
Optional External Temperature Probe Kit
VWR
®
Advanced Dry Block Heaters
VWR
®
DRY BLOCK HEATERS
VWR
®
Advanced Dry Block Heaters
Block Capacity
Dimensions (L x W x H)
Electrical (50/60 Hz)
Ship Weight
Cat. No.
1
12.4 x 8 x 3.5” (31.5 x 20.3 x 8.9cm)
120V | 0.9 amps | 110 watts
5.8lbs (2.6kg)
1
12.4 x 8 x 3.5” (31.5 x 20.3 x 8.9cm)
230V | 0.5 amps | 110 watts
5.8lbs (2.6kg)
2
15.4 x 8 x 3.5” (39.1 x 20.3 x 8.9cm)
120V | 1.7 amps | 210 watts
6.4lbs (2.9kg)
2
15.4 x 8 x 3.5” (39.1 x 20.3 x 8.9cm)
230V | 0.9 amps | 210 watts
6.4lbs (2.9kg)
3
18.4 x 8 x 3.5” (46.7 x 20.3 x 8.9cm)
120V | 1.7 amps | 210 watts
8lbs (3.6kg)
3
18.4 x 8 x 3.5” (46.7 x 20.3 x 8.9cm)
230V | 0.9 amps | 210 watts
8lbs (3.6kg)
4
16.9 x 8 x 3.5” (42.9 x 20.3 x 8.9cm)
120V | 2.6 amps | 310 watts
8.5lbs (3.9kg)
4
16.9 x 8 x 3.5” (42.9 x 20.3 x 8.9cm)
230V | 1.3 amps | 310 watts
8.5lbs (3.9kg)
6
20.9 x 8 x 3.5” (53.1 x 20.3 x 8.9cm)
120V | 3.4 amps | 410 watts
10lbs (4.5kg)
6
20.9 x 8 x 3.5” (53.1 x 20.3 x 8.9cm)
230V | 1.7 amps | 410 watts
10lbs (4.5kg)
1...,55,56,57,58,59,60,61,62,63,64 66,67,68,69,70,71,72,73,74,75,...116
Powered by FlippingBook