VWR Collection Laboratory Equipment - page 63
61
Support Plates
Designed to hold Hotplates/Stirrers and other apparatus to lab-frames or ring stands. Aluminum construction
offers strength and durability. Support plates include a non-skid rubber mat and can mount to vertical rods up
to 0.75” (1.9cm) diameter.
Support Rod and Clamp Kit
Designed for holding temperature probes, thermometers or other glass tubes. Includes an 18” (45.7cm)
stainless steel support rod, thermometer/temperature probe extension clamp, 3-prong medium swivel clamp,
and hook connector.
Recommended for all hotplates or hotplate-stirrers.
Description
Cat. No.
Support Rod and Clamp Kit
Size
Dimensions
Cat. No.
Small
9.5 x 6.5” (24.1 x 16.5cm)
Medium
12.75 x 9” (32.4 x 22.9cm)
Large
15.75 x 12” (40 x 30.5cm)
Hook Connector
3-Prong Swivel
Clamp
Extension Clamp
Support Rod and Clamp Kit Includes:
Small Support Plate recommended for Standard, Advanced 4 x 4 or Basic Mini Hotplates, Stirrers or Hotplate-
Stirrers.
Medium Support Plate recommended for Professional, Advanced, Standard 7 x 7 or Dyla Series Hotplates,
Stirrers or Hotplate-Stirrers.
Large Support Plate recommended for multiple units.
VWR
®
Supports
VWR
®
HOTPLATES/STIRRERS
1...,53,54,55,56,57,58,59,60,61,62 64,65,66,67,68,69,70,71,72,73,...116
Powered by FlippingBook