VWR Collection Laboratory Equipment - page 62
60
• Mixes up to 1500mL of water; easily stirs 100% glycerol at 20°C
• Safe, low voltage DC operation
• Electronic speed control
• 60 to 2000rpm stirring range
Compact, submersible magnetic stirrer is housed in a sealed, nickel-plated brass
case mounted on a solid brass base. A 38” (96.5cm) cable connects the power
supply to the stirring unit. Stirrer operates on low voltage DC and is safety fused
on DC side for safe operation underwater. Unit is easily mounted to rod supports
or laboratory frames with standard mounting bracket on rear of power supply. Can
also be used as a mini-stirrer in lattice work.
Ordering Information
Unit is supplied with a 40 x 8mm stir bar. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Model 700 Submersible Stirrer
120V | 0.16 amps | 20 watts
Model 700 Submersible Stirrer
230V | 0.08 amps | 20 watts
VWR
®
Submersible Stirrer
Submersible Stirrer
VWR
®
Dyla Series and Submersible Stirrer
VWR
®
HOTPLATES/STIRRERS
Specifications
Speed Range
60 to 2000rpm
Maximum Capacity
1500mL
Power
20 watts
Dimensions, Stirrer (Dia. x H)
2.75 x 2.75”
(7 x 7cm)
Dimensions, Power Supply (L x W x H)
5 x 4 x 3.5”
(12.7 x 10.2 x 8.9cm)
Ship Weight
2lbs (0.9kg)
• Large 6.5” diameter cast aluminum top plate
• Built-in support rod holder
The economical and durable VWR
®
Dyla Series hotplate, stirrer and hotplate-
stirrers are ideal for general heating and stirring. Large 6.5” (16.5cm) diameter
cast aluminum top plate provides a stable platform for large vessels. Even heating
surface is capable of boiling water or stirring with ease up to 1500mL of water.
Electronic speed control and bi-metallic thermostat delivers reliable and repeatable
temperature and speed control. Powerful heater reaches maximum temperature
in only minutes. Large magnet provides strong magnetic coupling to the spin bar.
Indicator lights illuminate when the heater is turned on. Built-in support rod holder
with locking knob accepts optional Support Rod and Clamp Kit. Optional Support
Rod and Clamp Kit allows unit to be mounted to lab-frames or ring stands (see
page 61).
Ordering Information
Unit is supplied with a 40 x 8mm stir bar. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Cat. No.
Dylatherm
®
Hotplate
120V | 10 amps | 575 watts
Dylatherm
®
Hotplate
230V | 5 amps | 575 watts
Dylastir
®
Stirrer
120V | 0.5 amps | 20 watts
Dylastir
®
Stirrer
230V | 0.25 amps | 20 watts
Dyla-Dual
®
Hotplate-Stirrer
120V | 10 amps | 575 watts
Dyla-Dual
®
Hotplate-Stirrer
230V | 5 amps | 575 watts
VWR
®
Dyla Series
Dyla Series
Specifications
Temperature Range
to 400°C
Speed Range
100 to 1200rpm
Maximum Capacity (H
2
O)
1500mL
Top Plate Material
Aluminum
Top Plate Dimensions (diameter)
6.5” (16.5cm)
Overall Dimensions (L x W x H)
8.75 x 8 x 4.75”
(22.2 x 20.3 x 12.1cm)
Ship Weight
4.6lb (2.1kg)
1...,52,53,54,55,56,57,58,59,60,61 63,64,65,66,67,68,69,70,71,72,...116
Powered by FlippingBook