VWR Collection Laboratory Equipment - page 61
59
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Positions Maximum Capacity (H
2
O) Maximum Vessel Diameter
Cat. No.
Standard Slow Speed Stirrer 1
120V | 1.3 amps | 75 watts
1
2L
5.9” (15cm)
Standard Slow Speed Stirrer 4
120V | 1.3 amps | 75 watts
4
1L
5.1” (13cm)
Standard Slow Speed Stirrer 4
230V | 1.1 amps | 90 watts
4
1L
5.1” (13cm)
VWR
®
Standard Slow Speed Stirrers
Description
Electrical (50/60 Hz)
Positions Maximum Capacity (H
2
O) Maximum Vessel Diameter
Cat. No.
Advanced Slow Speed Stirrer 1
120V | 1.3 amps | 75 watts
1
2L
5.9” (15cm)
Advanced Slow Speed Stirrer 4
120V | 1.3 amps | 75 watts
4
1L
5.1” (13cm)
Advanced Slow Speed Stirrer 4
230V | 1.1 amps | 90 watts
4
1L
5.1” (13cm)
Advanced Slow Speed Stirrer 10L 120V | 1.3 amps | 75 watts
1
10L
9.8” (25cm)
Advanced Slow Speed Stirrer 10L 230V | 1.1 amps | 90 watts
1
10L
9.8” (25cm)
VWR
®
Advanced Slow Speed Stirrers
VWR
®
Standard & Advanced Slow Speed Stirrers & Accessories
VWR
®
HOTPLATES/STIRRERS
Hotplate/Stirrer Accessories
Description
Cat. No.
8” (20.3 cm) Stainless Steel PT1000 Probe
8” (20.3 cm) PTFE PT1000 Probe
10” (25.4 cm) Stainless Steel PT1000 Probe
10” (25.4 cm) PTFE PT1000 Probe
Replacement Probe Kit with 8” (20.3cm) Stainless Steel PT1000 Probe
Temperature Probes
1...,51,52,53,54,55,56,57,58,59,60 62,63,64,65,66,67,68,69,70,71,...116
Powered by FlippingBook