VWR Collection Laboratory Equipment - page 59
57
Description
Electrical (50/60 Hz)
Positions Maximum Capacity (H
2
O) Maximum Vessel Diameter
Cat. No.
Standard Multi-Position Stirrer 4 120V | 1.3 amps | 85 watts
4
1L
5.1” (13cm)
Standard Multi-Position Stirrer 4 230V | 0.65 amps | 85 watts
4
1L
5.1” (13cm)
Standard Multi-Position Stirrer 5 120V | 1.3 amps | 85 watts
5
600mL
3.75” (9.5cm)
Standard Multi-Position Stirrer 5 230V | 0.65 amps | 85 watts
5
600mL
3.75” (9.5cm)
Standard Multi-Position Stirrer 6 120V | 1.3 amps | 85 watts
6
400mL
3.2” (8cm)
Standard Multi-Position Stirrer 6 230V | 0.65 amps | 85 watts
6
400mL
3.2” (8cm)
Standard Multi-Position Stirrer 9 120V | 1.3 amps | 85 watts
9
250mL
3” (7.5cm)
Standard Multi-Position Stirrer 9 230V | 0.65 amps | 85 watts
9
250mL
3” (7.5cm)
VWR
®
Standard Multi-Position Stirrers
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Units are also supplied with a 1” (2.5cm) PTFE
coated stir bar for each position. 2 year limited warranty on parts and labor.
Description
Electrical (50/60 Hz)
Positions Maximum Capacity (H
2
O) Maximum Vessel Diameter
Cat. No.
Advanced Multi-Position Stirrer 4 120V | 1.3 amps | 85 watts
4
1L
5.1” (13cm)
Advanced Multi-Position Stirrer 4 230V | 0.65 amps | 85 watts
4
1L
5.1” (13cm)
Advanced Multi-Position Stirrer 5 120V | 1.3 amps | 85 watts
5
600mL
3.75” (9.5cm)
Advanced Multi-Position Stirrer 5 230V | 0.65 amps | 85 watts
5
600mL
3.75” (9.5cm)
Advanced Multi-Position Stirrer 6 120V | 1.3 amps | 85 watts
6
400mL
3.2” (8cm)
Advanced Multi-Position Stirrer 6 230V | 0.65 amps | 85 watts
6
400mL
3.2” (8cm)
Advanced Multi-Position Stirrer 9 120V | 1.3 amps | 85 watts
9
250mL
3” (7.5cm)
Advanced Multi-Position Stirrer 9 230V | 0.65 amps | 85 watts
9
250mL
3” (7.5cm)
VWR
®
Advanced Multi-Position Stirrers
VWR
®
Standard & Advanced Multi-Position Stirrers
VWR
®
HOTPLATES/STIRRERS
1...,49,50,51,52,53,54,55,56,57,58 60,61,62,63,64,65,66,67,68,69,...116
Powered by FlippingBook