VWR Collection Laboratory Equipment - page 58
56
• Stirs up to 9 vessels at once
• Synchronized and uniform mixing
• Low profile design
VWR
®
Standard and Advanced Multi-Position Stirrers are powerful magnetic stirrers
with precise speed control that offers repeatable performance. Synchronized operation
ensures uniform mixing at each position.
Operating Features:
Low profile design:
Takes up minimal space. Spill-resistant housing channels
fluids away from internal components. Cast aluminum base offers durability and
added stability.
Glass filled nylon top plate:
Features a chemical resistant, reflective white top
plate surface that is easy to clean.
Large top plate:
Stirs up to 9 vessels at once with helpful position indicators.
Microprocessor control:
Precisely regulates speed through the entire range.
Stirring function, with continuous duty motor and magnet, maintains set speed even
under changing load or viscosity.
Adjustment knob:
Basic speed control knob with dial markings from 1 to 10
(Standard Series).
LED display:
Touch pad controls with easy-to-read, independent LED displays
for speed and time. Speed is adjustable in 10rpm increments. Timer will display
elapsed time or, when programmed to user defined limit, will shut off unit when time
reaches zero (Advanced Series).
Safety Features:
Speed ramping feature:
Slowly increases speed for improved safety and enhanced
coupling. Avoids splashing, improves spin bar control and provides excellent low
speed control.
Audible alarm:
In timed mode, alarmwill sound when time reaches zero. (Advanced
Series)
Operating Conditions:
Units can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Dissolution studies, media/reagent preparations, and titration studies.
VWR
®
Standard & Advanced Multi-Position Stirrers
VWR
®
HOTPLATES/STIRRERS
Specifications
Speed Range
60 to 1400rpm
Speed Stability
± 2%
Timer (Advanced Series)
1 second to 160 hours
Top Plate Material
Glass filled nylon
Top Plate Dimensions (L x W)
12.5 x 11” (31.8 x 27.9cm)
Overall Dimensions (L x W x H)
17 x 11 x 4”
(43.2 x 27.9 x 10.2cm)
Ship Weight
14.75lbs (6.7kg)
1...,48,49,50,51,52,53,54,55,56,57 59,60,61,62,63,64,65,66,67,68,...116
Powered by FlippingBook