VWR Collection Laboratory Equipment - page 57
55
• LED Displays for speed and time
• Durable, stainless steel construction
• Mixes up to 200L
VWR
®
Advanced Model 1000 and Model 2000 Large Capacity Stirrers are ideal for
high volume applications. The powerful magnetic drive is capable of mixing high
viscosity materials. Stainless steel base offers durability and added stability.
Operating Features:
Microprocessor control:
Precisely regulates speed through the entire range.
Stirring function, with continuous duty motor and magnet, maintains set speed even
under changing load or viscosity.
LED display:
Touch pad controls with easy-to-read, independent LED displays
for speed and time. Timer will display elapsed time or, when programmed to user
defined limit, will shut off unit when time reaches zero.
Safety Features:
Audible alarm:
In timed mode, alarm will sound when time reaches zero.
Speed ramping Feature:
Slowly increases speed for improved safety and
enhanced coupling. Avoids splashing, improves spin bar control and provides
excellent low speed control.
Operating Conditions:
Units can be run in conditions from 5 to 40°C (41 to 104°F), maximum 80% relative
humidity, non-condensing.
Applications:
Mixing viscous materials, polymers, and pilot scale.
Description
Electrical (50/60 Hz)
Cat. No.
Model 1000 Advanced Large Capacity Stirrer, 100L
120V | 1 amp | 41 watts
Model 1000 Advanced Large Capacity Stirrer, 100L
230V | 0.5 amps | 41 watts
Model 2000 Advanced Large Capacity Stirrer, 200L
120V | 1 amp | 82 watts
Model 2000 Advanced Large Capacity Stirrer, 200L
230V | 0.5 amps | 82 watts
VWR
®
Advanced Large Capacity Stirrer
Ordering Information
Units include a 92” (234cm) detachable, 3-wire cord and plug (230 volt units are supplied with Euro type plug). Model 1000 units are supplied with a 2.75” (7cm)
egg shaped PTFE coated stir bar, Model 2000 units are supplied with a 4” (10.2cm) PTFE coated stir bar with a pivot ring. 2 year limited warranty on parts and
labor.
VWR
®
Advanced Large Capacity Stirrers
VWR
®
HOTPLATES/STIRRERS
Specifications
Speed Range
100 to 1800rpm
(increased in 10rpm increments)
Speed Stability
± 2%
Timer
1 second to 160 hours
Maximum Capacity H
2
O
Model 1000
100L
Model 2000
200L
Maximum Weight Capacity
440lbs (193kg)
Top Plate Material
304 stainless steel
Top Plate Dimensions (L x W)
25 x 21.25” (63.5 x 54cm)
Overall Dimensions (L x W x H)
26.75 x 28.5 x 5.25”
(68 x 72.4 x 13.3cm)
Ship Weight
63lbs (28.6kg)
1...,47,48,49,50,51,52,53,54,55,56 58,59,60,61,62,63,64,65,66,67,...116
Powered by FlippingBook